Skip to Content
Merck
All Photos(1)

Documents

M3761

Supelco

Metronidazole

analytical standard

Synonym(s):

2-Methyl-5-nitroimidazole-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O3
CAS Number:
Molecular Weight:
171.15
Beilstein:
611683
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

crystalline

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

159-161 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria
parasites

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

Mode of action

DNA synthesis | interferes

storage temp.

2-8°C

SMILES string

CC1=NC=C([N+]([O-])=O)N1CCO

InChI

1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3

InChI key

VAOCPAMSLUNLGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metronidazole is a nitroimidazole drug, used particularly against infections caused by anaerobic bacteria and protozoa in humans and domestic animals.

Application

The analytical standard can be used as follows:
  • Cyclic voltametric determination of metronidazole at carbon paste electrode (CPE) in three different brands of metronidazole tablets
  • Simultaneous determination of metronidazole, diloxanide, spiramycin, and cliquinol in pharmaceutical dosage forms by reversed-phase high-performance liquid chromatography coupled to photodiode array detector (RP-HPLC-DAD)
  • Development of an electrochemical sensor based on polydopamine/carboxylic multi-walled carbon nano-tubes (MWCNTs-COOH) nanocomposites for the determination of metronidazole
  • Liquid chromatography–ion trap mass spectrometry (LC-MS/MS based analysis of metronidazole in water, sediment, and fish tissue samples
  • Determination of metronidazole using an electrochemical sensor based on multi-walled carbon nanotubes (MWCNTs) and chitosan-nickel complexes (CTS-Ni) in tablets, human urine, and serum samples

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alison L Huckenpahler et al.
Translational vision science & technology, 9(10), 18-18 (2020-09-29)
To observe and characterize cone degeneration and regeneration in a selective metronidazole-mediated ablation model of ultraviolet-sensitive (UV) cones in zebrafish using in vivo optical coherence tomography (OCT) imaging. Twenty-six sws1:nfsB-mCherry;sws2:eGFP zebrafish were imaged with OCT, treated with metronidazole to selectively
Christina M Surawicz et al.
The American journal of gastroenterology, 108(4), 478-498 (2013-02-27)
Clostridium difficile infection (CDI) is a leading cause of hospital-associated gastrointestinal illness and places a high burden on our health-care system. Patients with CDI typically have extended lengths-of-stay in hospitals, and CDI is a frequent cause of large hospital outbreaks
Marcos Sande-Melón et al.
Cell reports, 29(4), 1041-1054 (2019-10-24)
During heart regeneration in the zebrafish, fibrotic tissue is replaced by newly formed cardiomyocytes derived from preexisting ones. It is unclear whether the heart is composed of several cardiomyocyte populations bearing different capacity to replace lost myocardium. Here, using sox10
T H I Brummer et al.
BJOG : an international journal of obstetrics and gynaecology, 120(10), 1269-1276 (2013-06-22)
To evaluate cefuroxime and metronidazole antibiotic prophylaxis. Observational nonrandomised 1-year prospective cohort study. Fifty-three hospitals in Finland. A total of 5279 women undergoing hysterectomy for benign indications, with cefuroxime given to 4301 and metronidazole given to 2855. Excluding other antibiotics
Samrah Masud et al.
Autophagy, 15(5), 796-812 (2019-01-25)
Innate immune defense against intracellular pathogens, like Salmonella, relies heavily on the autophagy machinery of the host. This response is studied intensively in epithelial cells, the target of Salmonella during gastrointestinal infections. However, little is known of the role that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service