Skip to Content
Merck
All Photos(1)

Key Documents

D7008

Sigma-Aldrich

Dihydroethidium

≥95% purity, powder

Synonym(s):

2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine, 3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3
CAS Number:
Molecular Weight:
315.41
Beilstein:
5107482
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Dihydroethidium, ≥95%

Quality Level

Assay

≥95%

form

powder

technique(s)

titration: suitable

color

pink to very dark red

solubility

chloroform: 20 mg/mL

fluorescence

λex 370 nm; λem 420 nm (for cytoplasm of living cells)
λex 535 nm; λem 610 nm (for chromatin of living cells)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

CCN1C(c2ccccc2)c3cc(N)ccc3-c4ccc(N)cc14

InChI

1S/C21H21N3/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14/h3-13,21H,2,22-23H2,1H3

InChI key

XYJODUBPWNZLML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dihydroethidium has been used as a superoxide probe in bovine pulmonary artery endothelial cell (BPAEC). It has been used to check the in situ generation of ROS (reactive oxygen species).
Redox indicator. Blue fluorescence until oxidized to ethidium.

Packaging

Packaged under Argon.

Linkage

B-ring reduced analog of ethidium

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cira Rubies et al.
Scientific reports, 9(1), 11443-11443 (2019-08-09)
Obstructive sleep apnea syndrome (OSA) promotes aortic dilatation, increased stiffness and accelerated atherosclerosis, but the mechanisms of vascular remodelling are not known. We aimed to assess vascular remodelling, its mechanisms, and the effect of mesenchymal stem cells (MSC) infusions in
Quercetin reduces oxidative stress and inhibits activation of c-Jun N-terminal kinase/activator protein-1 signaling in an experimental mouse model of abdominal aortic aneurysm.
Wang L, et al.
Molecular Medicine Reports, 9, 435-442 (2014)
20-HETE increases superoxide production and activates NAPDH oxidase in pulmonary artery endothelial cells.
Medhora M, et al.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 294, L902-L911 (2008)
Rossarin Suwanarusk et al.
The Journal of antimicrobial chemotherapy, 70(1), 124-129 (2014-08-26)
Methylene blue, once discarded due to its unsettling yet mild side effects, has now found a renewed place in the pharmacopoeia of modern medicine. The continued spread of drug-resistant Plasmodium vivax and Plasmodium falciparum has also led to a recent
Karina A Roth et al.
Journal of endodontics, 38(9), 1253-1256 (2012-08-16)
Root canal fillings are intended to prevent microbial proliferation over time in the canal after treatment. The objective of this study was to assess biofilm proliferation within the sealer-dentin interfaces of 2 methacrylate resin-based systems, self-etch (SE) and total-etch (TE)

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service