Skip to Content
Merck
All Photos(2)

Key Documents

D2535

Sigma-Aldrich

Direct Blue 15

suitable for Histopaque® system, Powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H24N6O16S4Na4
CAS Number:
Molecular Weight:
992.80
Colour Index Number:
24400
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Direct Blue 15, suitable for Histopaque® system, suitable for viability studies of collagenase-treated rat liver cells

description

suitable for Histopaque® system

Quality Level

form

powder

composition

Dye content, ~44%

color

dark blue

solubility

water: 10 mg/mL, clear, blue

suitability

suitable for viability studies of collagenase-treated rat liver cells

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].COc1cc(ccc1N=Nc2c(O)c3c(N)cc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O)-c4ccc(N=Nc5c(O)c6c(N)cc(cc6cc5S([O-])(=O)=O)S([O-])(=O)=O)c(OC)c4

InChI

1S/C34H28N6O16S4.4Na/c1-55-25-9-15(3-5-23(25)37-39-31-27(59(49,50)51)11-17-7-19(57(43,44)45)13-21(35)29(17)33(31)41)16-4-6-24(26(10-16)56-2)38-40-32-28(60(52,53)54)12-18-8-20(58(46,47)48)14-22(36)30(18)34(32)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4

InChI key

OLSOUGWNONTDCK-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

May be used as a substitute for trypan blue for some biological and staining applications.

Legal Information

Histopaque is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzhong Zhan et al.
Journal of hazardous materials, 187(1-3), 348-354 (2011-01-29)
Copper hydroxide nitrate (Cu(2)(OH)(3)NO(3)) was synthesized solvothermally in anhydrous ethanol and characterized by XRD, FTIR, TG-DTA and SEM. The peroxide degradation of an azo dye (Direct Blue 15) on this material was evaluated by examining catalyst loading, initial pH, hydrogen
A Kai et al.
International journal of biological macromolecules, 20(3), 221-231 (1997-06-01)
The difference of influence of a certain kind of direct dye on the structure of nascent microbial cellulose was examined, with Direct Red 28 have a biphenylenebis(azo) skeletal structure; Direct Blue 1 having two hydroxyl, two methoxy and two sulfonate
Nurdan Kasikara Pazarlioglu et al.
Water environment research : a research publication of the Water Environment Federation, 82(7), 579-585 (2010-07-31)
To investigate biodegradability by Trametes versicolor, five structurally different direct azo-dyes--Direct Black 38, Direct Blue 15 (DB 15), Direct Orange 26, Direct Green 6, and Direct Yellow 12--were studied. The DB 15 was determined as the best biodegradable dye by
Samta Saroj et al.
Chemosphere, 107, 240-248 (2014-01-15)
Soils contaminated with dyes were collected and screened for obtaining potential fungal strains for the degradation of azo dyes. A strain that demonstrated broad spectrum ability for catabolizing different azo dyes viz. Acid Red 183 (AR 183), Direct Blue 15
F Rafii et al.
Applied and environmental microbiology, 59(6), 1731-1734 (1993-06-01)
The purified azoreductase and nitroreductase of Clostridium perfringens, which have similar electrophoretic properties, both reacted in a Western blot (immunoblot) with a polyclonal antibody raised against the azoreductase. The activity of both enzymes was enhanced by flavin adenine dinucleotide and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service