Skip to Content
Merck
All Photos(3)

Key Documents

C3143

Sigma-Aldrich

Chromotrope 2R

Powder

Synonym(s):

2-(Phenylazo)chromotropic acid disodium salt, Acid Red 29

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N2Na2O8S2
CAS Number:
Molecular Weight:
468.37
Colour Index Number:
16570
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Chromotrope 2R, suitable for modified Gomori Trichrome stain

form

powder

composition

Dye content, ≥70%

technique(s)

microbe id | staining: suitable

color

red to brown

solubility

H2O: 1 mg/mL, dark red

εmax

≥450 at 510-513 nm in methanol

suitability

suitable for modified Gomori Trichrome stain

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1cc(cc2cc(c(\N=N\c3ccccc3)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H12N2O8S2.2Na/c19-12-8-11(27(21,22)23)6-9-7-13(28(24,25)26)15(16(20)14(9)12)18-17-10-4-2-1-3-5-10;;/h1-8,19-20H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b18-17+;;

InChI key

GXEAXHYQKZAJGB-QIKYXUGXSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chromotrope 2R has been used in the Gomori trichrome staining in osteochondral tissues. It has also been used for the staining of eosinophils in lung sections.

Biochem/physiol Actions

Chromotrope 2R is used for the staining of microsporidia in body fluids and stool samples. It is used in modified Gomori′s trichrome staining. Chromotrope 2R has high affinity for phospholipids and is used for the staining of mitochondrial and endoplasmic reticulum membrane in histological samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antigen sensitization influences organophosphorus pesticide-induced airway hyperreactivity.
Proskocil BJ, et al.
Environmental Health Perspectives, 116, 381-381 (2008)
Bennett JE, et al.
Mandell, Douglas, and Bennett's Principles and Practice of Infectious Diseases (2014)
Chitosan-glycerol phosphate/blood implants increase cell recruitment, transient vascularization and subchondral bone remodeling in drilled cartilage defects.
Chevrier A, et al.
Osteoarthritis and Cartilage, 15, 316-316 (2007)
Eveline P C T de Rijk et al.
Toxicologic pathology, 31(4), 462-464 (2003-07-10)
Hyaline droplet formation in the proximal tubular cells of the kidney commonly occurs under different pathological conditions in experimental animals. In rats, intracytoplasmic hyaline droplet formation is predominantly associated with accumulation of the male rat-specific alpha(2u)-globulin, whereas under other (pathological)
L Cameron et al.
Journal of immunology (Baltimore, Md. : 1950), 164(3), 1538-1545 (2000-01-21)
Eosinophil differentiation occurs within the bone marrow in response to eosinopoietic cytokines, particularly IL-5. Recently, however, eosinophil precursors (CD34/IL-5Ralpha+ cells) and IL-5 mRNA+ cells have been identified within the lungs of asthmatics, indicating that a population of eosinophils may differentiate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service