Skip to Content
Merck
All Photos(1)

Key Documents

C0426000

Caprylic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Octanoic acid, Acid C8, Caprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
Beilstein:
1747180
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 78 °C)

API family

caprylic acid

manufacturer/tradename

EDQM

refractive index

n20/D 1.428 (lit.)

bp

237 °C (lit.)

mp

15-17 °C (lit.)

density

0.91 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCC(O)=O

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)

InChI key

WWZKQHOCKIZLMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Caprylic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romain Harmancey et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(10), 3966-3978 (2013-07-05)
Diabetic patients with acute myocardial infarction are more likely to die than nondiabetic patients. In the present study we examined the effect of insulin resistance on myocardial ischemia tolerance. Hearts of rats, rendered insulin resistant by high-sucrose feeding, were subjected
Hong-Fei Tong et al.
Journal of chromatography. A, 1285, 88-96 (2013-03-12)
Hydrophobic charge-induction chromatography (HCIC) is a novel downstream bioprocessing technology for antibody purification and it has several advantages over traditional purification processes. However, its separation selectivity still needs to be improved. In this work, sodium caprylate (NaCA) was used as
F Perri et al.
Italian journal of gastroenterology and hepatology, 30(2), 211-217 (1998-07-24)
Non-invasive evaluation of gastric emptying is generally performed by scintigraphy which is, however, not always applicable and is not suitable for children and child-bearing women. Recently, a new method based on stable isotope breath testing has been introduced in clinical
K M Morrill et al.
Journal of dairy science, 95(7), 3987-3996 (2012-06-23)
Our objectives were to evaluate the use of refractometry as a means of estimating immunoglobulin G (IgG) concentration of bovine maternal colostrum (MC) and determine if fractionation of MC using caprylic acid (CA) improved estimates of IgG. Samples (n=85) of
Marta Librán-Pérez et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 164(1), 241-248 (2012-09-27)
To assess the hypothesis that an acute dietary fatty acid (FA) supply may improve glucose tolerance in rainbow trout, we orally administered fish with fish oil (FO; 10mL.kg(-1), one time), which were then subjected to a glucose tolerance test and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service