Skip to Content
Merck
All Photos(1)

Key Documents

BCR701

Sediment (extractable elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

storage temp.

2-8°C

Application

Sediment (extractable elements) has been used as a standard reference material to assess the arsenic extraction efficiency of BCR sequential and single extraction methods from water sediment samples.

Analysis Note

For more information please see:
BCR701

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arsenic fractionation in sediments of different origins using BCR sequential and single extraction methods
Baig AJ, et al.
Journal of Hazardous Materials, 167, 745-751 (2009)
M Cyriac et al.
Marine pollution bulletin, 164, 112042-112042 (2021-02-02)
This study investigates the influence of upwelling induced seasonal hypoxia in the sediment-water interface on the distribution, bioavailability and geochemical partitioning of selected trace metals in surficial sediments along the southwest coast of India based on two successive cruises. The
Saranya Jayachandran et al.
Marine pollution bulletin, 133, 920-929 (2018-07-26)
Impact of pH variation of overlying water column on transport and transformation of Cu-sediment complexes in the bottom mangrove sediments was investigated by using different metal extraction studies. The total Cu concentration in the studied sediments varied from ~64 ± 1 to
Darwin Ramteke et al.
The Science of the total environment, 764, 143798-143798 (2021-01-09)
Geochemical fractionation study of cadmium (Cd) in combination with equilibrium based chemical speciation modelling (visual MINTEQ) in finer sediment samples (silt and clay portion), provided a better description of Cd bioavailability for sessile organisms (like oysters) in the Zuari estuary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service