Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8704

Buffalo river sediment

NIST® RM 8704

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

One unit of RM 8704 contains 50 grams of radiation-sterilized, freeze-dried, homogenized river sediment.

Application

Buffalo river sediment Reference material (RM) is designed for the analysis of soils, sediments, or materials with a similar matrix.

Features and Benefits

  • NIST certificate is provided with both reference and information values of elements mass fraction.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, drying, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Magnesium (Mg), Calcium (Ca), Potassium (K), Carbon (C), Sodium (Na), Iron (Fe) , Titanium (Ti), Antimony (Sb), Lead (Pb), Barium (Ba), Manganese (Mn), Cadmium (Cd), Nickel (Ni), Cerium (Ce), Scandium (Sc), Cesium (Cs), Thorium (Th), Chromium (Cr), Uranium (U), Cobalt (Co), Vanadium (V), Europium (Eu), Zinc (Zn), Hafnium (Hf)

Reference/Informational value is provided for the following analytes.
Arsenic (As)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service