Skip to Content
Merck
All Photos(1)

Key Documents

PHR1306

Supelco

3-tert-Butyl-4-hydroxyanisole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-tert-Butyl-4-methoxyphenol, 3-BHA, BHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O2
CAS Number:
Molecular Weight:
180.24
Beilstein:
1867499
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1083100

API family

bha

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1ccc(O)c(c1)C(C)(C)C

InChI

1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3

InChI key

MRBKEAMVRSLQPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
3-tert-Butyl-4-hydroxyanisole is a synthetic antioxidant, widely used as a food additive to prevent oxidative deterioration of fats and oils.[1]

Application

3-tert-Butyl-4-hydroxyanisole may be used as a pharmaceutical reference standard for the determination of the analyte in plasma samples using high-resolution capillary gas chromatography-mass spectrometry with selective ion monitoring.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0066 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the antioxidant 3-tert.-butyl-4-hydroxyanisole in rat plasma using high-resolution gas chromatography?mass spectrometry
Bailey E, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 225(1), 83-89 (1981)
Nathalie Allaman-Pillet et al.
Molecular cancer research : MCR, 13(1), 86-97 (2014-08-22)
Retinoblastoma is the most common pediatric intraocular neoplasm. While retinoblastoma development requires the inactivation of both alleles of the retinoblastoma tumor suppressor gene (RB1) in the developing retina, additional genomic changes are involved in tumor progression, which progressively lead to
Hye Jin Jung et al.
PloS one, 9(11), e112256-e112256 (2014-11-15)
Silver materials have been widely used in diverse fields. However, their toxicity and their mechanism, especially in different forms, have not been studied sufficiently. Thus, cytotoxicity, apoptosis, and interleukin-1beta (IL-1β) production were investigated using macrophage-like THP-1 cells in the presence
Huilin Liu et al.
Journal of agricultural and food chemistry, 63(9), 2545-2549 (2015-02-24)
A novel optosensing chip was constructed by anchoring the fluorescence sensing material layer based on molecularly imprinted polymer (MIP) coated CdSe/ZnS quantum dots (QDs) on a chip for highly selective and sensitive optosensing of sesamol in sesame oil. Many factors
Pedro Bullón et al.
Pharmacological research, 91, 1-8 (2014-12-03)
Oxidative stress is implicated in several infectious diseases. In this regard, lipopolysaccharide (LPS), an endotoxic component, induces mitochondrial dysfunction and oxidative stress in several pathological events such as periodontal disease or sepsis. In our experiments, LPS-treated fibroblasts provoked increased oxidative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service