Skip to Content
Merck
All Photos(1)

Key Documents

B1120000

Biperiden hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(5-Bicyclo[2.2.1]hept-2-enyl)-1-phenyl-3-(1-piperidinyl)propan-1-ol hydrochloride, Akineton, Akinophyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30ClNO
CAS Number:
Molecular Weight:
347.92
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

biperiden

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].OC(CCN1CCCCC1)(C2CC3CC2C=C3)c4ccccc4

InChI

1S/C21H29NO.ClH/c23-21(19-7-3-1-4-8-19,11-14-22-12-5-2-6-13-22)20-16-17-9-10-18(20)15-17;/h1,3-4,7-10,17-18,20,23H,2,5-6,11-16H2;1H

InChI key

RDNLAULGBSQZMP-UHFFFAOYSA-N

Gene Information

human ... CHRM1(1128)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Biperiden hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Antiparkinsonian; non-selective muscarinic receptor antagonist.
Biperiden hydrochloride is antiparkinsonian; non-selective muscarinic receptor antagonist. It is used for the adjunctive treatment of all forms of Parkinson′s Disease (postencephalitic, idiopathic, and arteriosclerotic); also commonly used to improve parkinsonian signs and symptoms related to antipsychotic drug therapy. LD50 in rats 750 mg/kg; in dogs 340 mg/kg.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service