Skip to Content
Merck
All Photos(1)

Key Documents

A8097

Sigma-Aldrich

Acridine Orange hydrochloride solution

>95.0% purity (HPLC), solution

Synonym(s):

3,6-Bis(dimethylamino)acridine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3 · HCl
CAS Number:
Molecular Weight:
301.81
Beilstein:
3575587
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)

Quality Level

Assay

≥95.0% (HPLC)

form

solution

concentration

10 mg/mL in H2O

technique(s)

microbe id | staining: suitable

color

orange to red

εmax

47-54 at 486-492 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].CN(C)c1ccc2cc3ccc(cc3nc2c1)N(C)C

InChI

1S/C17H19N3.ClH/c1-19(2)14-7-5-12-9-13-6-8-15(20(3)4)11-17(13)18-16(12)10-14;/h5-11H,1-4H3;1H

InChI key

VSTHNGLPHBTRMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acridine Orange hydrochloride solution has been used to study autophagic cell death. It has also been used for the staining of chromosomes.

Biochem/physiol Actions

Acridine Orange is a metachromatic dye which can stain DNA, RNA and acid glycosaminoglycans. At low concentration it intercalates into DNA and precipitates RNA. However, at high concentration it denatures and precipitates both RNA and DNA. Acridine Orange is also used to analyze autophagy. It goes into acidic organelles in a pH-dependent manner. At neutral pH, acridine orange gives a green fluorescence and in acidic conditions, it accumulates in the acidic organelle giving a bright red fluorescence.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nikolay A Pyataev et al.
Polymers, 11(5) (2019-05-28)
This study looked into the synthesis and study of Dextrane Sulfate-Doxorubicin Nanoparticles (DS-Dox NP) that are sensitive to amylase and show anticoagulant properties. The particles were obtained by the method of solvent replacement. They had a size of 305 ±
Gamze Guney Eskiler et al.
Journal of biochemical and molecular toxicology, 33(8), e22338-e22338 (2019-04-14)
The aim of this study was to investigate the combined effects of usnic acid (UA) and Tamoxifen (Tam) or Enzalutamide (Enz) on hormone receptor-positive breast and prostate cancer (BC and PC), respectively. The antiproliferative and apoptotic effects of Tam or
Murugesan Sathiya Deepika et al.
Materials science & engineering. C, Materials for biological applications, 103, 109716-109716 (2019-07-28)
Improved therapeutic effects can be achieved by the delivery of combination of drugs through multifunctional cell targeted nanocarrier systems. The present investigation reports the preparation of Poly (D,L-lactic-co-glycolic acid) (PLGA) nanospheres loaded with the novel combination such as Rutin (R)
Dung Thuy Nguyen Pham et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 50(3), 791-805 (2019-06-30)
Biofilm formation is one of the resistance mechanisms of Pseudomonas aeruginosa against antimicrobial compounds. Biofilm formation also characterizes for the infection and pathogenesis of P. aeruginosa, along with production of various virulence factors. With recent development of nanotechnology, the present
Yanhui Guan et al.
Cellular signalling, 62, 109339-109339 (2019-06-08)
Diabetes mellitus-related cardiomyopathy (DMCMP) has been defined as ventricular dysfunction that occurs in diabetic patients independent of a recognized cause such as coronary artery disease or hypertension. Mechanisms underlying DMCMP have not been fully elucidated. In this study, the roles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service