Skip to Content
Merck
All Photos(4)

Documents

158550

Sigma-Aldrich

Acridine Orange hemi(zinc chloride) salt

Dye content 85 %

Synonym(s):

3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Basic Orange 14

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H20ClN3 · HCl · 1/2ZnCl2
CAS Number:
Molecular Weight:
369.96
Colour Index Number:
46005
Beilstein:
3734978
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 85%

solubility

H2O: 1 mg/mL, clear, orange to very deep orange

λmax

489 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].Cl[H].Cl[Zn]Cl.CN(C)c1ccc2cc3ccc(cc3nc2c1)N(C)C.CN(C)c4ccc5cc6ccc(cc6nc5c4)N(C)C

InChI

1S/2C17H19N3.4ClH.Zn/c2*1-19(2)14-7-5-12-9-13-6-8-15(20(3)4)11-17(13)18-16(12)10-14;;;;;/h2*5-11H,1-4H3;4*1H;/q;;;;;;+2/p-2

InChI key

RAHGLSRJKRXOSY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Acridine Orange hemi (zinc chloride) salt has been used to stain primary human umbilical vein endothelial cells (HUVEC) and is used in micronucleus tests (MNTs).
Acridine Orange, a cell-permeable metachromatic fluorescent cationic dye that intercalates DNA and RNA, is used in fluorescence and epiflouresence microscopy. Acridine Orange dye has been used to analyze mitochondria and lysosomal content by flow cytometry, characterize multidrug resistance, and measure changes in mitochondrial mass during apoptosis in rat thymocytes.

Biochem/physiol Actions

Acridine orange (AO) is a fluorescent dye with two different staining characteristics. In fixed tissues, it acts as a metachromatic dye and differentially stains DNA (green) and RNA (orange). But, in living cells it acts as a pH indicator and produce brilliant orange-red in acidic condition. AO has been used to identify apoptotic cells in drosophila embryos. Acridine Orange staining method has been used to deliver more consistent data in the micronucleus test.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular chromosome breaks on silicon surface.
Jiang J
Biomaterials, 30(14), 2661-2665 (2009)
Labilizing action of intercalating drugs and dyes on bacterial ribosomes.
A D Wolfe et al.
Biochemistry, 11(9), 1569-1572 (1972-04-25)
An application of Acridine Orange fluorescent staining to the micronucleus test.
Hayashi M
Mutation Research, 120(4), 241-247 (1983)
Toxicity of waterborne Copper in premetamorphic tadpoles of Lithobates catesbeianus (Shaw, 1802).
Ossana NA
Bulletin of Environmental Contamination and Toxicology, 84(6), 712-715 (2010)
J F Golden et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 27(1), 522-528 (1979-01-01)
A system for screening cervical cytological preparations is described which employs the Leitz Texture Analyzer System (E. Leitz, Rockleigh, N. J.) quantitative staining with acridine orange, and a fluorescence standard. The instrumentation scans cells on microscope slides and detects objects

Related Content

Nancy-520 for DNA Detection and Quantitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service