A2908
Amino acid standards for protein hydrolysates containing norleucine
analytical standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
NACRES:
NA.24
Recommended Products
grade
analytical standard
analyte chemical class(es)
amino acids, peptides, proteins
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
food and beverages
format
neat
storage temp.
2-8°C
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
Amino acids listed are 0.5 μmole/ml except L-cystine at 0.25 μmole/ml in 0.2 N sodium citrate, pH 2.2.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
L I I Ouoba et al.
Journal of applied microbiology, 94(3), 396-402 (2003-02-18)
To examine isolates of Bacillus subtilis and B. pumilus predominant in Soumbala for their ability to degrade African locust bean proteins (ALBP). Agar diffusion test in casein and ALBP agar was used for screening of isolates. The profiles of water-soluble
Yael Politi et al.
Arthropod structure & development, 46(1), 30-38 (2016-06-23)
We identify the presence of multiple vascular channels within the spider fang. These channels seem to serve the transport of zinc to the tip of the fang to cross-link the protein matrix by binding to histidine residues. According to amino
Tina Ukmar-Godec et al.
Journal of structural biology, 192(3), 392-402 (2015-10-04)
The limpet radula is a feeding organ, which contains more than 100 rows of teeth. During their growth the teeth mature and advance in position along the radula. The simpler doccoglossan radulae operate by grinding rocky substrates, extracting the algae
W N U Perera et al.
British poultry science, 60(4), 404-413 (2019-04-19)
1. Two experiments were conducted to assess the nitrogen-corrected apparent metabolisable energy (AMEn; Exp. 1; 288 Ross 308 male broilers at d 14; 36 cages with eight birds each) and coefficient of standardised ileal digestibility (CSID) of amino acids (AA;
A Cowieson et al.
Poultry science, 98(9), 3722-3728 (2019-03-12)
A total of 144 male Ross 308 broiler chickens were used in a digestibility bioassay to determine the additivity of apparent or standardized amino acid (AA) digestibility values for corn, soybean meal (SBM), or a mixture of corn and SBM.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service