Skip to Content
Merck
All Photos(1)

Documents

A9781

Supelco

Amino acid standards for protein hydrolysates

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
eCl@ss:
32160406
NACRES:
NA.24

grade

analytical standard

composition

L-Alanine
Ammonium chloride
L-Arginine
L-Aspartic acid
L-Cystine
L-Glutamic acid
Glycine
L-Histidine
L-Isoleucine
L-Leucine
L-Lysine
L-Methionine
L-Phenylalanine
L-Proline
L-Serine
L-Threonine
L-Tryosine
L-Tryptophan
L-Valine

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

application(s)

food and beverages

format

neat

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Amino acids listed are 0.5 μmole/mL except L-cystine at 0.25 μmole/mL in 0.2 N sodium citrate, pH 2.2.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaolin Zhang et al.
Biotechnology progress, 36(4), e2975-e2975 (2020-02-06)
Media components play an important role in modulating cell metabolism and improving product titer in mammalian cell cultures. To sustain cell productivity, highly active oxidative metabolism is desired. Here we explored the effect of tricarboxylic acid (TCA) cycle intermediates supplementation
Amino acid analysis of mucins.
J X Yan et al.
Methods in molecular biology (Clifton, N.J.), 125, 113-119 (2000-05-23)
Marina Bacci et al.
Cell reports, 28(1), 104-118 (2019-07-04)
Endocrine therapy (ET) is the standard of care for estrogen receptor-positive (ER+) breast cancers. Despite its efficacy, ∼40% of women relapse with ET-resistant (ETR) disease. A global transcription analysis in ETR cells reveals a downregulation of the neutral and basic
Yu Zhong et al.
Journal of agricultural and food chemistry, 65(50), 10875-10883 (2017-11-29)
Microalgae were studied as function bioaccumulators of selenium (Se) for food and feed supplement. To investigate the bioaccumulation of Se and its effects on the unicellular green alga Chlorella pyrenoidosa, the algal growth curve, fluorescence parameters, antioxidant enzyme activity, and
Neil Templeton et al.
Metabolic engineering, 44, 126-133 (2017-09-28)
Industrial cell culture requires substantial energy to generate protein. The protein generated is not only the product of interest (IgG in this case), but also the protein associated with biomass. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service