Skip to Content
Merck
All Photos(1)

Key Documents

A1269000

L-Arabinitol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-(−)-Arabitol, L-Arabinitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O5
CAS Number:
Molecular Weight:
152.15
Beilstein:
1720521
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

arabinitol

manufacturer/tradename

EDQM

mp

101-104 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC[C@H](O)C(O)[C@@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4-/m0/s1

InChI key

HEBKCHPVOIAQTA-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

ʟ-Arabinitol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Teresa J Stradomska et al.
Journal of medical microbiology, 59(Pt 12), 1490-1496 (2010-08-21)
A non-invasive, non-culture-based method of determining urinary D-/L-arabinitol (D-/L-ARA) ratios was investigated as a tool for the diagnosis of invasive candidiasis in nosocomial paediatric infection cases. The study encompassed 138 children aged 4 days to 16 years (mean ± SD=1.6
Tomoyuki Toyoda et al.
Journal of industrial microbiology & biotechnology, 38(9), 1179-1185 (2010-11-18)
D-arabitol production from lactose by Kluyveromyces lactis NBRC 1903 has been studied by following the time courses of concentrations of cell mass, lactose, D: -arabitol, ethanol, and glycerol at different temperatures. It was found that temperature is a key factor
Renata Bura et al.
Journal of industrial microbiology & biotechnology, 39(7), 1003-1011 (2012-03-09)
An endophytic yeast, Rhodotorula mucilaginosa strain PTD3, that was isolated from stems of hybrid poplar was found to be capable of production of xylitol from xylose, of ethanol from glucose, galactose, and mannose, and of arabitol from arabinose. The utilization
Byoung Hoon Yoon et al.
Biotechnology letters, 33(4), 747-753 (2010-12-04)
Xylose reductase (XR) is a key enzyme in biological xylitol production, and most XRs have broad substrate specificities. During xylitol production from biomass hydrolysate, non-specific XRs can reduce L-arabinose, which is the second-most abundant hemicellulosic sugar, to the undesirable byproduct
Monika Kordowska-Wiater et al.
Polish journal of microbiology, 61(4), 291-297 (2012-01-01)
L-arabitol is used in the food and pharmaceutical industries. It can be secreted by genetically modified Saccharomyces cerevisiae carrying the genes responsible for pentose metabolism in yeast cells. The process of the biotransformation of L-arabinose to arabitol is highly dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service