Skip to Content
Merck
All Photos(1)

Key Documents

93645

Supelco

Dihexylammonium acetate solution

0.5 M in H2O, suitable for ion pair chromatography, LiChropur

Synonym(s):

Dihexylamine acetate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3(CH2)5]2NH · CH3COOH
CAS Number:
Molecular Weight:
245.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NB.21

form

liquid

Quality Level

quality

LiChropur

concentration

0.45-0.55 M
0.5 M in H2O

technique(s)

ion pair chromatography: suitable

refractive index

n20/D 1.350-1.354

density

0.992-0.996 g/mL at 20 °C

SMILES string

CC(O)=O.CCCCCCNCCCCCC

InChI

1S/C12H27N.C2H4O2/c1-3-5-7-9-11-13-12-10-8-6-4-2;1-2(3)4/h13H,3-12H2,1-2H3;1H3,(H,3,4)

InChI key

VAMSKEFYAOCCKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dihexylammonium acetate can be used as an ion-pairing agent for the analysis of nucleosides and nucleotides in foodstuffs and beverages, inositol phosphates in nutrition products, and acid orange dyes in meat products by ion-pair chromatography based techniques.

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of nucleosides and nucleotides in dietary foods and beverages using ion-pairing liquid chromatography-electrospray ionization-mass spectrometry
Yamaoka N, et al.
Journal of Chromatography. B, Biomedical Applications, 878(23), 2054-2060 (2010)
Fast Determination of Three Acid Orange Dyes: Acid Orange ?, Acid Orange ? and Acid Orange AGT in Meat Products by Ion-pair Ultra Performance Liquid Chromatography-Tandem Mass Spectrometry
ZHANG SF, et al.
Journal of Instrumental Analysis, 10 (2012)
Kohei Kuwada et al.
Journal of natural medicines, 74(1), 135-141 (2019-08-14)
LC-MS analyses of saponin fractions of Achyranthes roots in the Japanese market revealed that there were three patterns for the saponin fraction of their water extracts, i.e., the saponins with a sugar moiety at position 28 [achyranthosides B (3), C
Determination of nucleosides and nucleotides in food samples by using liquid chromatography and capillary electrophoresis
Dominguez-Alvarez J, et al.
TrAC, Trends in Analytical Chemistry, 92, 12-31 (2017)
Rapid method for simultaneous determination of inositol phosphates by IPC-ESI-MS/MS and its application in nutrition and genetic research
Zhang S, et al.
Chromatographia, 80(2), 275-286 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service