Skip to Content
Merck
All Photos(1)

Key Documents

329207

Sigma-Aldrich

Dianhydro-D-glucitol

98%

Synonym(s):

1,4:3,6-Dianhydro-D-sorbitol, D-Isosorbide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
Beilstein:
80510
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]20/D +45.2°, c = 3 in H2O

mp

60-63 °C (lit.)

SMILES string

O[C@H]1CO[C@@H]2[C@H](O)CO[C@H]12

InChI

1S/C6H10O4/c7-3-1-9-6-4(8)2-10-5(3)6/h3-8H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

KLDXJTOLSGUMSJ-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dianhydro-D-glucitol is a greener ingredient widely used as a monomer and building block for a variety of polymers and functional materials, for medical and pharmaceutical applications. It is also used as a fuel or fuel additive.
Applications include:
  • Preparation of renewable polyesters for coating applications.
  • Synthesis of isosorbide esters such as oligo(isosorbide adipate) (OSA), oligo(isosorbide suberate) (OSS), and isosorbide dihexanoate (SDH) as renewable alternatives to phthalate plasticizers.
  • Synthesis of biodegradable porous polyurethane scaffolds for tissue repair and regeneration.
  • Synthesis of the aspirin prodrug, isosorbide-2,5-diaspirinate (ISDA).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark T Ledwidge et al.
Atherosclerosis, 221(2), 478-483 (2012-02-14)
To evaluate the pharmacokinetics and effects of the first immediate-release (IR) niacin-aspirin prodrug (ST0702) on lipid, prostaglandin and thromboxane levels in non-human primates (NHPs). We compared 28 mg/kg crystalline IR niacin, equimolar doses of crystalline IR ST0702 and control on
Neal T Wasserman et al.
Veterinary ophthalmology, 16(1), 20-24 (2012-04-20)
To evaluate and compare the effects of oral isosorbide and glycerol on intraocular pressure (IOP), serum osmolality (SOSM), and blood glucose (BG) in normal dogs. Ten normal dogs were administered an oral dose of either isosorbide (1.5 g/kg), glycerol (1.5g/kg)
Jonathan Lai et al.
ChemSusChem, 4(8), 1104-1111 (2011-07-19)
The telomerisation of 1,3-butadiene with a Pd/TPPTS catalytic system in water or an organic solvent was used for the synthesis of C8 ethers from isosorbide, an agro-based diol. The use of water/oil biphasic reaction conditions allowed the selective synthesis of
Discovery of a ?true? aspirin prodrug.
Moriarty LM, et al.
Journal of Medicinal Chemistry, 51(24), 7991-7999 (2008)
Ali Hamidi Madani et al.
Urological research, 39(5), 361-365 (2011-01-06)
It has been suggested that nitrates are potent smooth muscle relaxants that may reduce pain and facilitate ureteral stone passage; therefore it may be an option for medical expulsive therapy in ureteral stones. In a prospective randomized controlled clinical trial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service