Skip to Content
Merck
All Photos(1)

Key Documents

90970

Supelco

γ-Butyrolactone

analytical standard

Synonym(s):

γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
Beilstein:
105248
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

851 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

16 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.436 (lit.)

bp

204-205 °C (lit.)

mp

−45 °C (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=C1CCCO1

InChI

1S/C4H6O2/c5-4-2-1-3-6-4/h1-3H2

InChI key

YEJRWHAVMIAJKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Butyrolactone (GBL) is a related lactone analog of γ-hydroxybutyric acid (GHB), a widespread drug of abuse, identified as an endogenous constituent of the mammalian brain, in which it exhibits a strong central nervous system depressant effect. GBL is considered as a legal substitute of GHB, and commonly identified in beverages including coffee, apple cider vinegar, tea, carbonated drinks, etc.

Application

γ-Butyrolactone may be used as an analytical reference standard for the quantification of the analyte in roasted Italian chestnuts, wine samples, beverages using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Precursor of γ-hydroxybutyric acid (GHB). It blocks dopamine release by blocking impulse flow in dopaminergic neurons. Pretreatment with γ-butyrolactone allows detection of autoreceptor-induced dopamine release.
Precursor of γ-hydroxybutyric acid (GHB); blocks dopamine release.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sensory evaluation of black instant coffee beverage with some volatile compounds present in aromatic oil from roasted coffee
Oliveira LA, et al.
Food Sci. Technol., 29(1), 76-80 (2009)
Polydimethylsiloxane solid-phase microextraction--gas chromatography method for the analysis of volatile compounds in wines: its application to the characterization of varietal wines
Pozo-Bayon AM, et al.
Journal of Chromatography A, 922(1-2), 267-275 (2001)
Micellar electrokinetic chromatographic screening method for common sexual assault drugs administered in beverages
Bishop CS, et al.
Forensic Science International, 141(1), 7-15 (2004)
Simultaneous Quantification of $\gamma$-Hydroxybutyrate, $\gamma$-Butyrolactone, and 1, 4-Butanediol in Four Kinds of Beverages
Jin S, et al.
International Journal of Analytical Chemistry, 2020(1), 7-15 (2020)
Volatile compound analysis of SPME headspace and extract samples from roasted Italian chestnuts (Castanea sativa Mill.) using GC-MS
Krist S, et al.
European Food Research and Technology, 219(5), 470-473 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service