Skip to Content
Merck
All Photos(1)

Key Documents

87920

Supelco

Tetramethylsilane

analytical standard, for NMR spectroscopy, ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(CH3)4
CAS Number:
Molecular Weight:
88.22
Beilstein:
1696908
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

ACS reagent
analytical standard
for NMR spectroscopy

Quality Level

vapor pressure

11.66 psi ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

842 °F

refractive index

n20/D 1.358 (lit.)
n20/D 1.359

bp

26-28 °C (lit.)

mp

−99 °C (lit.)

density

0.648 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C

InChI

1S/C4H12Si/c1-5(2,3)4/h1-4H3

InChI key

CZDYPVPMEAXLPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetramethylsilane has been recommended by the International Union of Pure and Applied Chemistry (IUPAC) as a universal reference for all nuclides. It is commonly used as a reference standard in nuclear magnetic resonance (NMR) for measuring proton chemical shifts and the temperature dependence of the 1H chemical shift of TMS in solvents such as chloroform, methanol, and dimethylsulfoxide is studied.[1]

Application

Tetramethylsilane may be used as an internal standard for the quantitative analysis of medicinal plant extracts and herbal products using the quantitative NMR (qNMR) method.[2] It may also be used as an internal standard to investigate peroxide-based chemical systems for the crosslinking reactions carried on isotactic polypropylene using FT-IR spectrometry and proton nuclear magnetic resonance (1H NMR) techniques.[3]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temperature dependence of the 1H chemical shift of tetramethylsilane in chloroform, methanol, and dimethylsulfoxide
Hoffman ER, et al.
Journal of Magnetic Resonance, 176(1), 87-98 (2005)
Monitoring the chemical crosslinking of propylene polymers through rheology
Romani F, et al.
Polymer, 43(4), 1115-1131 (2002)
Siddheshwar K Chauthe et al.
Phytochemical analysis : PCA, 23(6), 689-696 (2012-06-19)
Quantitative analysis and standardisation of plant extracts or herbal products is a tedious process requiring time-consuming sample preparation and analytical method development for the resolution of analyte peaks from the complex natural extract. Quantitative analysis by HPLC requires a pure
Thierry Rohmer et al.
The journal of physical chemistry. A, 115(42), 11696-11714 (2011-08-19)
The open-chain tetrapyrrole compound bilirubin was investigated in chloroform and dimethyl sulfoxide solutions by liquid-state NMR and as solid by (1)H, (13)C, and (15)N magic-angle spinning (MAS) solid-state NMR spectroscopy. Density functional theory (DFT) calculations were performed to interpret the
Jose C Pichardo et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 52(9), 1482-1489 (2011-07-30)
The current gold standard for measuring marrow cellularity is the bone marrow (BM) biopsy of the iliac crest. This measure is not predictive of total marrow cellularity, because the biopsy volume is typically small and fat fraction varies across the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service