Skip to Content
Merck
All Photos(1)

Documents

269913

Supelco

3-(Trimethylsilyl)propionic-2,2,3,3-d4 acid sodium salt

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCD2CD2CO2Na
CAS Number:
Molecular Weight:
172.27
Beilstein:
4046239
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

isotopic purity

98 atom % D

Assay

≥98.0% (NMR)

impurities

≤0.5% water (Karl Fischer)

pH

7.0-9.0 (1% in solution)

mass shift

M+4

SMILES string

[Na+].[2H]C([2H])(C([O-])=O)C([2H])([2H])[Si](C)(C)C

InChI

1S/C6H14O2Si.Na/c1-9(2,3)5-4-6(7)8;/h4-5H2,1-3H3,(H,7,8);/q;+1/p-1/i4D2,5D2;

InChI key

OIIWPAYIXDCDNL-HGFPCDIYSA-M

Application


  • Rapid separation and quantitation of curcuminoids combining pseudo two-dimensional liquid flash chromatography and NMR spectroscopy: This research demonstrates the application of 3-(Trimethylsilyl)propionic-2,2,3,3-d₄ acid sodium salt in the field of NMR spectroscopy, particularly emphasizing its role as an internal standard in the quantification of curcuminoids. The study showcases a novel method that combines pseudo two-dimensional liquid flash chromatography with NMR spectroscopy, providing a faster, more accurate analytical approach. This method significantly enhances the efficiency and reliability of chemical analysis in pharmaceutical and biomedical research, making it a valuable technique for analytical chemists in academia (Jayaprakasha et al., 2013).


Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harri Koskela et al.
Magnetic resonance in chemistry : MRC, 55(10), 917-927 (2017-04-30)
The spectral parameters of selected nerve agent degradation products relevant to the Chemical Weapons Convention, namely, ethyl methylphosphonate, isopropyl methylphosphonate, pinacolyl methylphosphonate and methylphosphonic acid, were studied in wide range of pH conditions and selected temperatures. The pH and temperature
Qiongju Cao et al.
Food & function, 11(4), 3095-3104 (2020-03-21)
Soups show diverse health functions, which could be linked to their original nutrient profiles and metabolites derived from digestion. NMR spectroscopy is a robust and rapid method that unveils or identifies the chemical composition of food or food-derived metabolites. In
Aline L Baptista et al.
JBMR plus, 4(7), e10372-e10372 (2020-07-16)
Bone biopsy is still the gold standard to assess bone turnover (T), mineralization (M), and volume (V) in CKD patients, and serum biomarkers are not able to replace histomorphometry. Recently, metabolomics has emerged as a new technique that could allow
Ekaterina Nevedomskaya et al.
Metabolomics : Official journal of the Metabolomic Society, 8(6), 1227-1235 (2012-11-09)
Urinary tract infection (UTI) encompasses a variety of clinical syndromes ranging from mild to life-threatening conditions. As such, it represents an interesting model for the development of an analytically based scoring system of disease severity and/or host response. Here we
Sung Ha Ryu et al.
The Journal of toxicological sciences, 44(1), 57-71 (2019-01-11)
Cisplatin (CP) is an anti-cancer drug used for treatment of solid tumors, but the major adverse effect is drug-induced nephrotoxicity. The current study aimed to determine biomarkers that might predict nephrotoxicity induced by CP using serum or urinary proton nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service