Skip to Content
Merck
All Photos(1)

Key Documents

86616

Supelco

Tetraethylammonium chloride

Tetraethylammonium chloride

for electrochemical analysis, ≥99.0%

Synonym(s):

TEA chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Cl)
CAS Number:
Molecular Weight:
165.70
Beilstein:
3563247
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Pricing and availability is not currently available.

grade

for electrochemical analysis

Quality Level

Assay

≥99.0% (AT)
≥99.0%

form

crystals

solubility

methanol: 0.1 g/mL, clear, colorless

SMILES string

[Cl-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.ClH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

YMBCJWGVCUEGHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fernanda S Gravina et al.
PloS one, 9(10), e111307-e111307 (2014-10-23)
The pacemaker mechanisms activating phasic contractions of vaginal and cervical smooth muscle remain poorly understood. Here, we investigate properties of pacemaking in vaginal and cervical tissues by determining whether: 1) functional pacemaking is dependent on the phase of the estrus
Daniel Ulrich
The European journal of neuroscience, 40(4), 2600-2607 (2014-06-04)
The thalamic reticular nucleus (nRt) is an assembly of GABAergic projection neurons that participate in the generation of brain rhythms during synchronous sleep and absence epilepsy. NRt cells receive inhibitory and excitatory synaptic inputs, and are endowed with an intricate
Min Shen et al.
Vascular pharmacology, 58(1-2), 64-70 (2012-07-24)
Evidence has indicated that resveratrol (Res) produces vasorelaxation and may decrease the coronary heart disease mortality. However, several pathways involved in the mechanism of vasorelaxation are still unclear. This study was designed, therefore, to test the probable ion channels or
Nanomechanics of cation-π interactions in aqueous solution.
Qingye Lu et al.
Angewandte Chemie (International ed. in English), 52(14), 3944-3948 (2013-03-02)
Ling Xiao et al.
Circulation research, 112(10), 1310-1322 (2013-03-28)
A chromosomal haplotype producing cardiac overexpression of dipeptidyl peptidase-like protein-6 (DPP6) causes familial idiopathic ventricular fibrillation. The molecular basis of transient outward current (I(to)) in Purkinje fibers (PFs) is poorly understood. We hypothesized that DPP6 contributes to PF I(to) and

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service