Skip to Content
Merck
All Photos(4)

Key Documents

A9187

Sigma-Aldrich

Adenosine 5′-triphosphate magnesium salt

≥95%, bacterial

Synonym(s):

ATP, ATP magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N5O13P3 · xMg2+
CAS Number:
Molecular Weight:
507.18 (free acid basis)
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

bacterial

Quality Level

Assay

≥95%

form

powder

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

[Mg].Nc1ncnc2n(cnc12)C3OC(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)C(O)C3O

InChI

1S/C10H16N5O13P3.Mg.2H/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;

InChI key

CPGAIACWYFBIFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-triphosphate magnesium salt has been used:
  • as a supplement in intracellular medium for Ca2+ retention assay and fluorescence wide field imaging analysis
  • as a component in potassium methane sulfonate-based internal solution for in vitro electrophysiology of cortical neurons
  • as a component in action potential intracellular solution

Biochem/physiol Actions

Adenosine 5γ-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is use in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps. ATP serves as a coenzyme in a wide array of enzymatic reactions.
P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun Jong Kim et al.
European journal of pharmacology, 791, 686-695 (2016-10-30)
Pyrazole derivatives were originally suggested as selective blockers of the transient receptor potential cation 3 (TRPC3) and channel. In particular, pyr3 and 10 selectively inhibit TRPC3, whereas pyr2 (BTP2) and 6 inhibit ORAI1. However, their effects on background K+ channel
Comprehensive multilevel in vivo and in vitro analysis of heart rate fluctuations in mice by ECG telemetry and electrophysiology.
Fenske S, et al.
Nature Protocols, 11(1), 61-61 (2016)
A subpopulation of striatal neurons mediates Levodopa-induced dyskinesia.
Girasole AE, et al.
Neuron, 97(4), 787-795 (2018)
Spatial separation of mitochondrial calcium uptake and extrusion for energy-efficient mitochondrial calcium signaling in the heart.
De La Fuente S, et al.
Cell reports, 24(12), 3099-3107 (2018)
Lea Wagmann et al.
Archives of toxicology, 92(9), 2875-2884 (2018-08-08)
Transporter-mediated drug-drug interactions (DDI) may induce adverse clinical events. As drugs of abuse (DOA) are marketed without preclinical safety studies, only very limited information about interplay with membrane transporters are available. Therefore, 13 DOA of various classes were tested for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service