Skip to Content
Merck
All Photos(1)

Key Documents

80165

Supelco

Phytane

analytical standard

Synonym(s):

2,6,10,14-Tetramethylhexadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H42
CAS Number:
Molecular Weight:
282.55
Beilstein:
1744639
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

69-71 °C/0.001 mmHg (lit.)

density

0.791 g/mL at 20 °C (lit.)

application(s)

environmental
food and beverages

format

neat

SMILES string

CCC(C)CCCC(C)CCCC(C)CCCC(C)C

InChI

1S/C20H42/c1-7-18(4)12-9-14-20(6)16-10-15-19(5)13-8-11-17(2)3/h17-20H,7-16H2,1-6H3

InChI key

GGYKPYDKXLHNTI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carin von Mühlen et al.
Journal of chromatography. A, 1105(1-2), 39-50 (2005-10-04)
This article discusses the application of comprehensive two-dimensional gas chromatography (GC x GC) to samples derived from petrochemicals. The use of GC x GC for characterization of petroleum and petroleum derivatives, through group type analysis, such as benzene, toluene, ethylbenzene
T W Custer et al.
Environmental toxicology and chemistry, 20(3), 624-631 (2001-05-15)
Tree swallow (Tachycineta bicolor) and house wren (Troglodytes aedon) eggs and chicks were collected near a refinery site on the North Platte River, Casper. Wyoming, USA and at a reference site 10 km upstream. Total polycyclic aromatic hydrocarbon (PAH) concentrations
B Stallwood et al.
Journal of applied microbiology, 99(4), 794-802 (2005-09-16)
To identify native Antarctic bacteria capable of oil degradation at low temperatures. Oil contaminated and pristine soils from Signy Island (South Orkney Islands, Antarctica) were examined for bacteria capable of oil degradation at low temperatures. Of the 300 isolates cultured
M A Caja et al.
Die Naturwissenschaften, 95(11), 1073-1077 (2008-07-12)
Although turbidite deposits are classically considered to be good reservoir rocks for oil and gas, there are no reports concerning their source rock potential in the literature. The sediments from the Vallfogona Formation in the South-Eastern Pyrenees present numerous organic
I Schifter et al.
Marine pollution bulletin, 123(1-2), 165-174 (2017-09-11)
Vertical profiles of deep-water fluorescence determined by the chlorophyll sensor, polycyclic aromatic hydrocarbons, biomarkers, and other miscellaneous parameters measured in the southern Gulf of Mexico are reported. In the course of the survey, unexpected deep fluorescences were recorded (>1100m depth)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service