Skip to Content
Merck
All Photos(3)

Key Documents

P2870

Sigma-Aldrich

Pristane

synthetic, ≥98% (GC)

Synonym(s):

Norphytane, 2,6,10,14-Tetramethylpentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)(CH2)3CH(CH3)2
CAS Number:
Molecular Weight:
268.52
Beilstein:
1720538
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Quality Level

Assay

≥98% (GC)

form

liquid

mol wt

268.5

storage condition

(tightly closed)

color

colorless

refractive index

n20/D 1.438 (lit.)

bp

68 °C/0.001 mmHg (lit.)

solubility

hexane: miscible 10 % (v/v)

density

0.783 g/mL at 20 °C (lit.)

storage temp.

room temp

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC(C)C

InChI

1S/C19H40/c1-16(2)10-7-12-18(5)14-9-15-19(6)13-8-11-17(3)4/h16-19H,7-15H2,1-6H3

InChI key

XOJVVFBFDXDTEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pristane, also known as norphytane, is a saturated norterpenoid alkane derived from phytane. Its immunological properties are valuable for various applications, serving as an inflammation-inducing agent in tumorigenesis and autoimmune disease induction, facilitating research on conditions like rheumatoid arthritis and lupus. Pristane′s diverse applications span acting as an immunological adjuvant, inducing autoimmune diseases, facilitating monoclonal antibody production, and serving as an energy source, making it a valuable asset in biochemical research, cell culture, and molecular biology.

Application

Pristane also acts as an immunological adjuvant for generating monoclonal antibodies in mouse ascites and finds use as a biological marker. Additionally, pristane can also be used as the primary carbon and energy source for the growth of a bacterial strain, coryneform soil isolate. Its distinctive ability to induce plasmacytomas is particularly valuable for studying B-cell tumors like Burkitt′s lymphoma.
It is commonly used for priming animals, leading to granulomatous inflammation and disruption of local lymphatic drainage, resulting in increased ascitic yields. It has been used to induce systemic lupus in mice
Pristane has been used as an inflammation-inducing agent, leading to myeloma development in BALB/c strain mice. It has been used as a rheumatoid arthritis-inducing agent in rats.

Features and Benefits

High quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marli L Moraes et al.
Journal of nanoscience and nanotechnology, 19(7), 3772-3776 (2019-02-16)
The diagnostics of the autoimmune hemolytic anemia (AIHA), a rare disease caused by autoantibody-induced hemolysis, is still prone to false positives for it is based on visual observation in the so-called Direct Coombs test. In this study, we developed a
Animal Models of Rheumatoid Arthritis (I): Pristane-Induced Arthritis in the Rat.
Tuncel J
PLoS ONE, 11, e0155936-e0155936 (2016)
Zhenlu Chong et al.
Cellular & molecular immunology, 15(12), 1057-1070 (2018-10-03)
Systemic lupus erythematosus (SLE) is a systemic autoimmune disease, and the pathogenesis of SLE has not been fully elucidated. The E3 ubiquitin ligase FBXW7 has been well characterized in cancer as a tumor suppressor that can promote the ubiquitination and
Neutrophil-Derived IL-6 Potentially Drives Ferroptosis Resistance in B Cells in Lupus Kidney
Wang Z
Mediators of Inflammation, 2023, 9810733-9810733 (2023)
Acid-mediated reactions under microfluidic conditions: a new strategy for practical synthesis of biofunctional natural products.
Tanaka K and Fukase K
Beilstein Journal of Organic Chemistry, 5, 40-40 (2009)

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service