Skip to Content
Merck
All Photos(1)

Key Documents

79609

Sigma-Aldrich

Phosphorus pentoxide

puriss. p.a., ACS reagent, ≥98.0% (T)

Synonym(s):

Phosphoric anhydride, Phosphorus(V) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
P2O5
CAS Number:
Molecular Weight:
141.94
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 384 °C)
10 mmHg ( 238 °C)

Assay

≥98.0% (T)

form

powder or crystals

impurities

≤0.02% phosphorus trioxide (P2O3)

pH

1.5 (20 °C, 10 g/L)

mp

340 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤5 mg/kg

cation traces

As: ≤100 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤10 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

InChI

1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12

InChI key

YWEUIGNSBFLMFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorus pentoxide (P2O5) is a phosphorous oxide. It participates in Pictet-Gams synthesis. Phosphorus pentoxide supported on alumina has been used for solvent-free and microwave-assisted synthesis of 1, 5-benzodiazepine derivatives.

Application

Phosphorus pentoxide may be used in the synthesis of dichlorine heptoxide (Cl2O7).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 215-215 (1994)
Jae-Hyung Ahn et al.
Microbial ecology, 68(2), 271-283 (2014-04-01)
The effects of water-saving irrigation on emissions of greenhouse gases and soil prokaryotic communities were investigated in an experimental rice field. The water layer was kept at 1-2 cm in the water-saving (WS) irrigation treatment and at 6 cm in
Radek Pohl et al.
Organic & biomolecular chemistry, 13(16), 4693-4705 (2015-03-24)
Systematic structure-activity studies on a diverse family of nucleoside phosphonic acids has led to the development of potent antiviral drugs such as HPMPC (CidofovirTM), PMEA (AdefovirTM), and PMPA (TenofovirTM), which are used in the treatment of CMV-induced retinitis, hepatitis B
Alumina/phosphorus pentoxide (APP) as an efficient reagent for the synthesis of 1, 5-benzodiazepines under microwave irradiation.
Kaboudin B and Navaee K.
Heterocycles, 1443-1446 (2001)
Eagleson M.
Concise Encyclopedia Chemistry, 836-836 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service