Skip to Content
Merck
All Photos(1)

Key Documents

75712

Sigma-Aldrich

Diethyl oxalate

purum, ≥99.0% (GC)

Synonym(s):

Diethyl ethanedioate, Ethyl oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCOOC2H5
CAS Number:
Molecular Weight:
146.14
Beilstein:
606350
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.03 (vs air)

Quality Level

vapor pressure

1 mmHg ( 47 °C)

grade

purum

Assay

≥99.0% (GC)

form

liquid

refractive index

n20/D 1.410 (lit.)
n20/D 1.410

bp

185 °C (lit.)

mp

−41 °C (lit.)

density

1.076 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(=O)OCC

InChI

1S/C6H10O4/c1-3-9-5(7)6(8)10-4-2/h3-4H2,1-2H3

InChI key

WYACBZDAHNBPPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hina Zafar et al.
Journal of photochemistry and photobiology. B, Biology, 142, 8-19 (2014-12-10)
A condensation reaction between 1,2-diphenylethane-1,2-dione dihydrazone (DPEDDH) and dimethyl or diethyloxalate in methanol resulted in a novel Schiff base octaazamacrocyclic ligand, (L): (6,7,14,15-tetraoxa-2,3,10,11-tetraphenyl-1,4,5,8,9,12,13,16-octaazacyclohexadecane-1,3,9,11-tetraene). Subsequently metal complexes of the type [MLX2] and [CuL]X2; (M=Mn(II), Co(II), Ni(II) and Zn(II); X=Cl or NO3)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service