Skip to Content
Merck
All Photos(1)

Documents

73081

Supelco

Tetrachlorobisphenol A

analytical standard

Synonym(s):

4,4′-Isopropylidenebis(2,6-dichlorophenol), 2,2′,6,6′-Tetrachloro-4,4′-isopropylidenediphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C[C6H2(Cl)2OH]2
CAS Number:
Molecular Weight:
366.07
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

134-136 °C (lit.)

format

neat

SMILES string

CC(C)(c1cc(Cl)c(O)c(Cl)c1)c2cc(Cl)c(O)c(Cl)c2

InChI

1S/C15H12Cl4O2/c1-15(2,7-3-9(16)13(20)10(17)4-7)8-5-11(18)14(21)12(19)6-8/h3-6,20-21H,1-2H3

InChI key

KYPYTERUKNKOLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrachlorobisphenol A is classified under the family of bisphenols, which are extensively utilized in food and beverages as well as in the processing and packaging industries.

Application

Tetrachlorobisphenol A may be used as an analytical standard for the determination of the analyte in environmental water as well as sediments and sludge samples, food and beverage products, and dairy products by various chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of seven bisphenols in environmental water and solid samples by liquid chromatography-electrospray tandem mass spectrometry
Yang Y, et al.
Journal of Chromatography A, 1328, 26-34 (2014)
Xingwang Hou et al.
Environmental science & technology, 53(15), 8805-8812 (2019-07-10)
Tetrabromobisphenol A (TBBPA) is the most widely used brominated flame retardant (BFR), and it bioaccumulates throughout the food chains. Its fate in the first trophic level, plants, is of special interest. In this study, a four-day hydroponic exposure of TBBPA
A high-throughput screening method of bisphenols, bisphenols digycidyl ethers and their derivatives in dairy products by ultra-high performance liquid chromatography-tandem mass spectrometry
Cheng Y, et al.
Analytica Chimica Acta, 950(46), 98-107 (2017)
Luciano Molognoni et al.
Journal of agricultural and food chemistry, 67(9), 2691-2699 (2019-02-13)
A fast, less expensive, analytical approach with high metrologic reliability was developed to assist an official program for 21 marine biotoxins, monitoring in bivalve mollusks. The simultaneous analysis of lipophilic and hydrophilic marine biotoxins was achieved using a sample preparation
Huiming Cao et al.
Archives of toxicology, 91(12), 3897-3912 (2017-06-16)
Certain bisphenols (BPs) have been regarded as endocrine-disrupting chemicals due to their structural similarities to bisphenol A (BPA), a well-known weak estrogenic chemical. However, very limited data are currently available on the relationship between estrogenic activity and the structure of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service