Skip to Content
Merck
All Photos(1)

Documents

450502

Sigma-Aldrich

4,4′-Isopropylidenebis(2,6-dimethylphenol)

98%

Synonym(s):

Bisphenol A, tetramethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C[C6H2(CH3)2OH]2
CAS Number:
Molecular Weight:
284.39
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

mp

162-165 °C (lit.)

SMILES string

Cc1cc(cc(C)c1O)C(C)(C)c2cc(C)c(O)c(C)c2

InChI

1S/C19H24O2/c1-11-7-15(8-12(2)17(11)20)19(5,6)16-9-13(3)18(21)14(4)10-16/h7-10,20-21H,1-6H3

InChI key

ODJUOZPKKHIEOZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel Deye et al.
Bioorganic & medicinal chemistry, 17(3), 1353-1360 (2009-01-02)
A public compound library with 260,000 compounds was screened virtually by computational docking for novel inhibitors of the transmembrane enzyme sarco/endoplasmic reticulum calcium ATPase (SERCA). Docking was performed with the program GOLD in conjunction with a high resolution X-ray crystal
Christopher Elam et al.
European journal of medicinal chemistry, 46(5), 1512-1523 (2011-03-01)
Two screening protocols based on recursive partitioning and computational ligand docking methodologies, respectively, were employed for virtual screens of a compound library with 345,000 entries for novel inhibitors of the enzyme sarco/endoplasmic reticulum calcium ATPase (SERCA), a potential target for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service