Skip to Content
Merck
All Photos(1)

Key Documents

72545

Supelco

Nitrate ion standard solution

0.01 M NO3-, for ion-selective electrodes

Synonym(s):

Potassium nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
grade:
for ion-selective electrodes
form:
solution

grade

for ion-selective electrodes

Quality Level

form

solution

shelf life

limited shelf life, expiry date on the label

concentration

0.01 M NO3-

density

1.00 g/mL at 20 °C

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Nitrate ion standard solution may be used for the preparation of ion-selective membranes and electrodes used in the evaluation of soil macronutrients for testing the soil fertility.

Preparation Note

prepared with KNO3 and H2O

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of nitrate and potassium ion-selective membranes for soil macronutrient sensing.
Kim H J, et al.
Transactions of the ASABE, 49(3), 597-606 (2006)
Redrothu Hanumantharao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 99, 181-188 (2012-10-16)
Nonlinear optical crystals of Bis (l-glutamine) potassium nitrate (BGPN) were grown by slow evaporation technique at ambient temperature. Solubility and metastable zone width of BGPN in aqueous solution were determined. The grown crystal was characterized by single crystal XRD, Powder
Jaclyn Marie Hill et al.
Water research, 46(11), 3553-3562 (2012-04-25)
The excessive addition of nitrogen to watersheds is recognized as one of the main causes of the global deterioration of aquatic ecosystems and an increasing number of studies have shown that δ¹⁵N signatures of macrophytes may reflect the N-loading of
Wolfgang R Engelsberger et al.
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here
M Bahra et al.
Nitric oxide : biology and chemistry, 26(4), 197-202 (2012-01-31)
Ingestion of inorganic nitrate elevates blood and tissue levels of nitrite via bioconversion in the entero-salivary circulation. Nitrite is converted to NO in the circulation, and it is this phenomenon that is thought to underlie the beneficial effects of inorganic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service