Skip to Content
Merck
All Photos(1)

Key Documents

71642

Sigma-Aldrich

Sodium phosphate dibasic

purum p.a., anhydrous, ≥98.0% (T)

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

grade

purum p.a.

Assay

≥98.0% (T)

form

powder

loss

≤2% loss on drying, 110 °C

pH

8.8-9.4 (25 °C, 50 mg/mL in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

water: soluble 50 mg/mL at 25 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤200 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Effects of a commercial whitening toothpaste containing hydrogen peroxide and citric acid on dentin abrasion and erosion.: This study evaluates the effects of sodium phosphate dibasic in a commercial whitening toothpaste formulation. The research highlights its role in mitigating dentin abrasion and erosion, emphasizing its importance in dental care applications (Kim JH et al., 2023).

  • Electrospraying of Bio-Based Chitosan Microcapsules Using Novel Mixed Cross-Linker: Experimental and Response Surface Methodology Optimization.: Sodium phosphate dibasic is utilized in the cross-linking process of chitosan microcapsules via electrospraying. This study demonstrates its significance in optimizing the physical properties and stability of bio-based microcapsules for potential applications in drug delivery systems (Uko L et al., 2022).

  • Compound treatment of thiolated citrus high-methoxyl pectin and sodium phosphate dibasic anhydrous improved gluten network structure.: This research investigates the combined effect of sodium phosphate dibasic and thiolated citrus high-methoxyl pectin on improving the gluten network structure in food systems, highlighting its role in enhancing food texture and quality (Chen J et al., 2023).

  • Bioremediation of trichloroethylene-polluted groundwater using emulsified castor oil for slow carbon release and acidification control.: Sodium phosphate dibasic is used to control acidification in the bioremediation of trichloroethylene-polluted groundwater, demonstrating its application in environmental engineering and pollution control (Chen WT et al., 2022).

  • Use of cellulose nanofibril (CNF)/silver nanoparticles (AgNPs) composite in salt hydrate phase change material for efficient thermal energy storage.: The study utilizes sodium phosphate dibasic in the development of a composite material for thermal energy storage, showcasing its importance in enhancing the thermal properties and efficiency of phase change materials (Shen Z et al., 2021).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan A Rees et al.
Journal of proteome research, 14(1), 120-132 (2014-10-21)
Interactions between a host and a bacterial pathogen are mediated by cross-talk between molecules present on, or secreted by, pathogens and host binding-molecules. Identifying proteins involved at this interface would provide substantial insights into this interaction. Although numerous studies have
C Tony Liu et al.
Journal of the American Chemical Society, 136(29), 10349-10360 (2014-07-01)
Electrostatic interactions play an important role in enzyme catalysis by guiding ligand binding and facilitating chemical reactions. These electrostatic interactions are modulated by conformational changes occurring over the catalytic cycle. Herein, the changes in active site electrostatic microenvironments are examined
Samuel Adler et al.
Gastrointestinal endoscopy, 76(6), 1170-1174 (2012-10-03)
Colon capsule endoscopy (CCE) is a noninvasive technique for the detection of colorectal lesions. However, for CCE to be offered as an out-of-clinic procedure, the system needs to automatically alert the patient when to ingest the laxative (booster). We tested
Johanna Troberg et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 107-118 (2014-10-11)
Understanding drug glucuronidation in the dog, a preclinical animal, is important but currently poorly characterized at the level of individual enzymes. We have constructed cDNAs for the 10 dog UDP-glucuronosyltransferases of subfamily 1A (dUGT1As), expressed them in insect cells, and
Nahid Amini et al.
Journal of pharmaceutical and biomedical analysis, 98, 140-143 (2014-06-13)
Positron emission tomography (PET) is an imaging technique based on the use of radioligands labeled with short lived radionuclides, such as (11)C (t½=20.4min) and (18)F (t½=109.8min), which as a consequence often requires rapid plasma protein binding analysis methods. In addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service