Skip to Content
Merck
All Photos(1)

Key Documents

675415

Sigma-Aldrich

Acetonitrile

≥99.9%, HPLC Plus, suitable for HPLC, poly-coated bottles

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC Plus
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)

Product Name

Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles

grade

HPLC Plus

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

525 °F
973 °F

expl. lim.

16 %

technique(s)

HPLC: suitable

impurities

≤0.01% water

evapn. residue

≤0.0002%

color

colorless

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

solubility

water: soluble

density

0.786 g/mL at 25 °C (lit.)

absorption

≤1 mAU at 210 nm

UV absorption

λ: 195 nm Amax: ≤0.10
λ: 200 nm Amax: ≤0.02
λ: 228 nm Amax: ≤0.005

application(s)

food and beverages

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile (MeCN), an aliphatic nitrile, is a colorless liquid with a pleasant odor. It is widely used as a solvent and intermediate in organic syntheses. It is transparent to UV-visible light making it highly applicable in spectrophotometric and fluorimetric techniques. As MeCN has low viscosity, high elution strength and is highly miscible in water, it is utilized as a mobile phase component in many chromatographic techniques. Its infrared spectrum has been recorded. Synthesis of alkyl hydroperoxides in MeCN by alkane oxidation with hydrogen peroxide in the presence of iron complexes has been studied. The hydrogenation of MeCN to form ethylamine using Co–B amorphous alloy catalyst has been investigated.

Application


  • Anthocyanin Separation: Acetonitrile is employed in high-performance liquid chromatography (HPLC) for the selective separation of anthocyanins. Studies have demonstrated the benefits of replacing acetonitrile with methanol in the mobile phase to control the selectivity and improve the resolution of anthocyanin separation, providing a more sustainable approach to chromatographic analysis (Deineka et al., Journal of Analytical Chemistry, 2021).

  • Detection of α-Amino Acids: Acetonitrile is used in the development of chemiluminescence methods for detecting α-amino acids. A mixed solution of water, acetonitrile, and ethyl acetate has been shown to enhance the sensitivity and accuracy of amino acid detection, making it a valuable tool for biochemical and clinical analyses (Kan et al., American Journal of Analytical Chemistry, 2020).

  • Electrochemical Detection of Biomarkers: Acetonitrile is utilized in the electrochemical detection of biomarkers such as 5-formyluracil. Labeling with (2-benzimidazolyl) acetonitrile enhances the selectivity and sensitivity of the detection, making it an important method for biomedical research and diagnostics (Tang et al., Analytical Chemistry, 2021).

  • Ligand-Assisted Excitation of Europium Complexes: Acetonitrile is used in studying the ligand-assisted excitation of Eu(III) complexes. The interaction of europium complexes with xenon difluoride in acetonitrile solution provides insights into their photophysical properties, which are essential for developing luminescent materials for various applications, including lighting and display technologies (Masyagutova et al., Journal of Fluorine Chemistry, 2024).


Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid phase hydrogenation of acetonitrile to ethylamine over the Co-B amorphous alloy catalyst.
Li H, et al.
J. Catal., 214(1), 15-25 (2003)
Hydrogen peroxide oxygenation of alkanes including methane and ethane catalyzed by iron complexes in acetonitrile.
Shul'pin GB, et al.
Advanced Synthesis & Catalysis, 346(2-3), 317-332 (2004)
Infrared spectra of acetonitrile and acetonitrile-d3.
Pace EL and Noe LJ.
J. Chem. Phys. , 49, 5317-5325 (1968)
Acetonitrile, the polarity chameleon.
Zarzycki PK, et al.
Analytical and Bioanalytical Chemistry, 397(3), 905-908 (2010)
Eagleson M.
Concise Encyclopedia Chemistry, 6-6 (1994)

Related Content

Ibuprofen and impurity separation using Ascentis Express 90Å C18 column with excellent peak shape and low quantification limits.

Ibuprofen and impurity separation using Ascentis Express 90Å C18 column with excellent peak shape and low quantification limits.

Ibuprofen and impurity separation using Ascentis Express 90Å C18 column with excellent peak shape and low quantification limits.

Ibuprofen and impurity separation using Ascentis Express 90Å C18 column with excellent peak shape and low quantification limits.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service