Skip to Content
Merck
All Photos(1)

Key Documents

AX0156

Supelco

Acetonitrile

≥99.9% (GC), suitable for LC/MS, OmniSolv®

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12352117
EC Index Number:
200-835-2
technique(s):
HPLC: suitable
LC/MS: suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Acetonitrile, OmniSolv® LC-MS

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 8327
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019

Quality Level

reg. compliance

suitable for FDA C-010.02

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

HPLC: suitable
LC/MS: suitable

impurities

≤0.02% Water (H2O)
≤0.5 μeq/g Titrable base
≤1 μeq/g Titrable acid
≤50 ppb p-nitrophenol (LC/MS)
≤50 ppb reserpine (LC/MS)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

UV cutoff

≤190 nm

UV absorption

λ: 200 nm Amax: ≤0.1
λ: 210 nm Amax: ≤0.04
λ: 220 nm Amax: ≤0.02
λ: 230 nm Amax: ≤0.01
λ: 240 nm Amax: ≤0.01
λ: 250 nm Amax: ≤0.01
λ: 270 nm Amax: ≤0.01
λ: 400-280 nm Amax: ≤0.005

SMILES string

CC#N

format

neat

shipped in

ambient

storage temp.

room temp

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile is a water-soluble polar organic solvent with exceptional solubilizing power, unique chromatographic properties, and ideal spectroscopic characteristics. [1]

Application

Acetonitrile is used as a:
  • Mobile phase solvent in reversed-phase chromatography. [1]
  • Rinsing solvent for Solid-phase extraction (SPE) column. [2]

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minColor (APHA)10APHA maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence as quinine at 254 nm1ppb maxFluorescence as quinine at 365 nm1ppb maxFormClear liquidGradient at 210 nm2mAU maxGradient at 254 nm0.3mAU maxIdentity (IR-spectrum)ConformsLC-MS purity, ESI(+), as reserpine50ppb maxLC-MS purity, ESI(-), as p-nitrophenol50ppb maxMetals, suitable for LC/MSTo pass testTitrable acid1µeq/g maxTitrable base0.5µeq/g maxUV Abs. at 200 nm0.1AU maxUV Abs. at 210 nm0.04AU maxUV Abs. at 220 nm0.02AU maxUV Abs. at 230 nm0.01AU maxUV Abs. at 240 nm0.01AU maxUV Abs. at 250 nm0.01AU maxUV Abs. at 270 nm0.01AU maxUV Abs. at 400-280 nm0.005AU maxUV Cut-off190nm maxWater (H2O)0.02% max

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of the sensitivity of evaporative universal detectors and LC/MS in the HILIC and the reversed-phase HPLC modes.
Clifford M R, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 877, 4133-4139 (2009)
High-performance liquid chromatographic procedure for the determination of a non-nucleoside HIV-1 reverse transcriptase inhibitor in human plasma.
E W, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 577, 129-134 (1992)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service