Skip to Content
Merck
All Photos(1)

Key Documents

59975

Supelco

Jasmone

analytical standard

Synonym(s):

cis-3-Methyl-2-(2-pentenyl)-2-cyclopenten-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
Beilstein:
1907713
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of isomers, GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

~6% trans-isomer

refractive index

n20/D 1.498 (lit.)
n20/D 1.500

bp

134-135 °C/12 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]\C(CC)=C(/[H])CC1=C(C)CCC1=O

InChI

1S/C11H16O/c1-3-4-5-6-10-9(2)7-8-11(10)12/h4-5H,3,6-8H2,1-2H3/b5-4-

InChI key

XMLSXPIVAXONDL-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: mentha

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Pawełczyk et al.
European journal of medicinal chemistry, 41(5), 586-591 (2006-03-28)
cis-Jasmone, from jasmonoid group, is an important jasmine odor fragrance compound. The syntheses of new heterocyclic analogues of jasmone were described. Five analogues of this compound were prepared under microwave irradiation and the results of the microwave assisted syntheses were
Stefan Dötterl et al.
Journal of chemical ecology, 38(12), 1539-1543 (2012-11-13)
Many plants attract their pollinators with floral scents, and these olfactory signals are especially important at night, when visual signals become inefficient. Dynastid scarab beetles are a speciose group of night-active pollinators, and several plants pollinated by these insects have
Anna Pawełczyk et al.
European journal of medicinal chemistry, 44(7), 3032-3039 (2008-09-06)
Taking the rising interest in jasmone structure based fragrant compounds into account it has been decided to take up an attempt to synthesize the new heterocyclic derivatives of this 2,3-disubstituted cyclopentenone, which could be characterized by the ability of interaction
Laxmi Yeruva et al.
Experimental lung research, 32(10), 499-516 (2006-12-16)
The jasmonates, cis-jasmone (CJ) and methyl jasmonate (MJ), were investigated for their effects against NSCLC cell lines A549 and H520. CJ or MJ inhibited the proliferation of both cell lines in a dose-dependent manner as well as induced cell cycle
A M El-Sayed et al.
Journal of chemical ecology, 35(6), 656-663 (2009-05-16)
This work was undertaken to identify floral compound(s) produced by honeysuckle flowers, Lonicera japonica (Thunberg), that mediate the attraction of New Zealand flower thrips Thrips obscuratus (Crawford). Volatiles were collected during the day and night and analyzed by gas chromatography-mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service