Skip to Content
Merck
All Photos(2)

Key Documents

55016

Supelco

Theaflavin

analytical standard

Synonym(s):

3,4,5-Trihydroxy-1,8-bis[(2R,3R)-3,5,7-trihydroxy-2-chromanyl]-6-benzo[7]annulenone, 3,4,6-Trihydroxy-1,8-bis(3α,5,7-trihydroxy-2α-chromanyl)-5H-benzocyclohepten-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H24O12
CAS Number:
Molecular Weight:
564.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1C3=Cc4c(cc(O)c(O)c4C(=O)C(O)=C3)[C@H]5Oc6cc(O)cc(O)c6C[C@H]5O

InChI

1S/C29H24O12/c30-11-3-17(32)15-8-21(36)28(40-23(15)5-11)10-1-13-14(7-20(35)27(39)25(13)26(38)19(34)2-10)29-22(37)9-16-18(33)4-12(31)6-24(16)41-29/h1-7,21-22,28-33,35-37,39H,8-9H2,(H,34,38)/t21-,22-,28-,29-/m1/s1

InChI key

IPMYMEWFZKHGAX-ZKSIBHASSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Theaflavin is a polyphenol, naturally available in tea, which can mainly play a role in the inhibition of xanthine oxidase enzyme to yield uric acid. It can also find applications in reducing oxidative stress and can also act like scavengers of superoxide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annadurai Anandhan et al.
Brain research, 1433, 104-113 (2011-12-06)
Parkinson's disease (PD) is a progressive neurodegenerative disorder, characterized by loss of dopominergic neurons in substantia nigra pars compacta, and can be experimentally induced by the neurotoxin 1-methyl-4-phenyl-1, 2, 3, 6-tetrahydropyridine (MPTP). Chronic administration of MPTP/probenecid (MPTP/p) leads to oxidative
Mian Zu et al.
Antiviral research, 94(3), 217-224 (2012-04-24)
The theaflavins fraction (TF80%, with a purity of 80%) and three theaflavin (TF) derivatives from black tea have been found to exhibit potent inhibitory effects against influenza virus in vitro. They were evaluated with a neuraminidase (NA) activity assay, a
Gerlinde Grelle et al.
Biochemistry, 50(49), 10624-10636 (2011-11-08)
Causal therapeutic approaches for amyloid diseases such as Alzheimer's and Parkinson's disease targeting toxic amyloid oligomers or fibrils are still emerging. Here, we show that theaflavins (TF1, TF2a, TF2b, and TF3), the main polyphenolic components found in fermented black tea
Wenxiang Wang et al.
Journal of cardiovascular pharmacology, 59(5), 434-440 (2012-01-06)
A model of homocysteine-induced injury in vascular endothelial cells was established to evaluate the protective role of theaflavins on homocysteine-injured human vascular endothelial cells (HUVECs). The cells were co-incubated with 3 concentrations of theaflavins (5, 10, or 20 mg/L) and
Inhibition of xanthine oxidase and suppression of intracellular reactive oxygen species in HL-60 cells by theaflavin-3, 3 `-digallate,(?)-epigallocatechin-3-gallate, and propyl gallate
Lin K-J, et al.
Journal of Agricultural and Food Chemistry, 48, 2736-2743 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service