Skip to Content
Merck
All Photos(1)

Key Documents

46794

Supelco

Sulfadimethoxine

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O4S
CAS Number:
Molecular Weight:
310.33
Beilstein:
306856
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

clinical testing

format

neat

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

COc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC)n1

InChI

1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ZZORFUFYDOWNEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Sulfadimethoxine is an long acting sulfonamide antibiotic, which finds applications in pen-held lobsters, Homarus americanus for treatment of gaffkemia.[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfadimethoxine may be used as a reference standard in the determination of sulfadimethoxine in eggs using high performance liquid chromatography (HPLC).[3]

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid high-performance liquid chromatographic determining technique of sulfamonomethoxine, sulfadimethoxine, and sulfaquinoxaline in eggs without use of organic solvents.
Analytica Chimica Acta null
Photochemical fate of sulfadimethoxine in aquaculture waters.
Farusava N.
Environmental Science & Technology, 481(2), 255-259 (2003)
Disposition of sulfadimethoxine in the lobster (Homarus americanus).
OMJ and GMB
Veterinary and Human Toxicology, 30, 36-40 (1988)
Hiroichi Ozaki et al.
Veterinary microbiology, 150(1-2), 132-139 (2011-01-15)
To investigate the effects of rearing practices of commercial broiler chickens on the incidence of antimicrobial resistance in commensal Escherichia coli isolates, fecal E. coli isolates obtained in 4 farms were screened for anitimicrobial resistance. Ten E. coli isolates were
Ailiang Chen et al.
Biosensors & bioelectronics, 42, 419-425 (2012-12-12)
Quickly and sensitively detection of antibiotic residues in animal products often requires time-consuming techniques and expensive instrumentation. To address these limitations, a high sensitive aptasensor for sulfadimethoxine (SDM) using unmodified gold nanoparticles (AuNPs) was developed in the present study. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service