Skip to Content
Merck
All Photos(1)

Documents

31738

Supelco

Sulfapyridine

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-[2-pyridyl]benzene sulfonamide, N1-(Pyridin-2-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N3O2S
CAS Number:
Molecular Weight:
249.29
Beilstein:
222065
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2ccccn2

InChI

1S/C11H11N3O2S/c12-9-4-6-10(7-5-9)17(15,16)14-11-3-1-2-8-13-11/h1-8H,12H2,(H,13,14)

InChI key

GECHUMIMRBOMGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Sulfapyridine is a metabolite of Sulfasalazine. It is an antibiotic and is mostly used in treating rheumatoid arthritis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfapyridine (SPY) has been used as internal standard in HPLC method for determination of sulfonamides residue from honey samples.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A reliable high-performance liquid chromatography with ultraviolet detection for the determination of sulfonamides in honey.
Granja RH
Analytica Chimica Acta, 613(1), 116-119 (2008)
Fátima Fernández et al.
Biosensors & bioelectronics, 26(4), 1231-1238 (2010-07-20)
Techniques for immunosensing like surface plasmon resonance (SPR) may respond to the need for rapid screening methods to improve food safety. This paper describes the development of a novel portable six channel SPR biosensor based on the plasmon of gold
Hai Haham et al.
Environmental science & technology, 46(21), 11870-11877 (2012-10-02)
Sorption-desorption behavior of sulfapyridine was studied with three distinct soil types low in organic carbon with or without the introduction of exogenous dissolved organic matter (DOM). Experiments with bulk soils yielded sorption coefficients equivalent to those obtained with soils richer
Hanna Isaksson et al.
Physiology & behavior, 105(3), 877-884 (2011-11-09)
Whole grain rye products have previously been shown to increase feelings of satiety for up to 8h after intake under standardized conditions. This study was set out to investigate the sustainability of the satiating effect after regular consumption of breakfast
Albert Redo-Sanchez et al.
The Analyst, 136(8), 1733-1738 (2011-02-22)
We report the use of terahertz (THz) spectroscopy to explore the spectral properties of eleven antibiotics commonly used in livestock production. Eight of the eleven antibiotics showed specific fingerprints in the frequency range between 0.1 and 2 THz. The main

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service