Skip to Content
Merck
All Photos(1)

Key Documents

46035

Supelco

Bromadiolon

PESTANAL®, analytical standard

Synonym(s):

Bromodiolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H23BrO4
CAS Number:
Molecular Weight:
527.41
Beilstein:
1335336
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(CC(c1ccccc1)C2=C(O)c3ccccc3OC2=O)c4ccc(cc4)-c5ccc(Br)cc5

InChI

1S/C30H23BrO4/c31-23-16-14-20(15-17-23)19-10-12-22(13-11-19)26(32)18-25(21-6-2-1-3-7-21)28-29(33)24-8-4-5-9-27(24)35-30(28)34/h1-17,25-26,32-33H,18H2

InChI key

OWNRRUFOJXFKCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromadiolon is an anticoagulant rodenticide, widely used for controlling rodent population.

Application

Bromadiolon may be used as an analytical reference standard for the quantification of the analyte in biological matrices using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of bromadiolone in whole blood by high-performance liquid chromatography coupled with electrospray ionization tandem mass spectrometry
Jin C-M, et al.
Forensic Science International, 171(1), 52-56 (2007)
The use of bromadiolone to potentiate self-mutilatory bleeding: a case report.
Michael B Centorino et al.
Psychosomatics, 53(5), 489-493 (2012-06-05)
Mickaël Sage et al.
Environmental research, 110(7), 664-674 (2010-08-10)
In many countries, the fox (Vulpes vulpes), predator of small mammals, is particularly affected by anticoagulant rodenticides such as bromadiolone due to secondary poisoning. Nevertheless, to date, no method of exposure monitoring is applicable in the field over large areas
[Bromadiolone poisoning complicated with thrombosis in the lower extremities in a case].
Jie Wu et al.
Zhonghua er ke za zhi = Chinese journal of pediatrics, 48(7), 550-551 (2010-11-09)
Julie Vein et al.
Ecotoxicology (London, England), 20(6), 1432-1441 (2011-06-02)
The anti-vitamin Ks (AVKs) are widely used to control rodent populations. They inhibit Vitamin K regeneration by the Vitamin K Epoxide Reductase (VKOR) and cause a fatal hemorrhagic syndrome. Because of repeated use, some populations of commensal rodents have expressed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service