Skip to Content
Merck
All Photos(1)

Key Documents

32677

Supelco

Difenacoum

PESTANAL®, analytical standard

Synonym(s):

3-(3-Biphenyl-4-yl-1,2,3,4-tetrahydro-1-naphthyl)-4-hydroxycoumarin, 3-(3-Biphenyl-4-yl-1,2,3,4-tetrahydro-naphthalen-1-yl)-4-hydroxychromen-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H24O3
CAS Number:
Molecular Weight:
444.52
Beilstein:
8360065
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC1=C(C2CC(Cc3ccccc23)c4ccc(cc4)-c5ccccc5)C(=O)Oc6ccccc16

InChI

1S/C31H24O3/c32-30-26-12-6-7-13-28(26)34-31(33)29(30)27-19-24(18-23-10-4-5-11-25(23)27)22-16-14-21(15-17-22)20-8-2-1-3-9-20/h1-17,24,27,32H,18-19H2

InChI key

FVQITOLOYMWVFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Difenacoum is a coumarin anticoagulant, which is developed as a rodenticide, in order to control warfarin‐resistant rodents.

Application

Difenacoum may be used as a reference standard in the determination of difenacoum in tissue samples using high performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M J Kelly et al.
Journal of chromatography, 620(1), 105-112 (1993-10-22)
A rapid and sensitive high-performance liquid chromatographic method for the analysis of cis and trans diastereomers of the anticoagulant rodenticide difenacoum has been described. The methodology demonstrates potential for the analysis of diastereomers of related 4-hydroxycoumarin anticoagulants. Separations were achieved
Field trials of second-generation anticoagulants against difenacoum-resistant Norway rat population.
Greaves HJ, et al.
Epidem. Inf., 89(2), 295-301 (1982)
R Wallin
The Biochemical journal, 236(3), 685-693 (1986-06-15)
In the liver, it appears that there are two different pathways for vitamin K reduction. One pathway is irreversibly inhibited by coumarin anticoagulant drugs. The other pathway has been shown in the present study to be composed of enzymes that
S Terneu et al.
Acta clinica Belgica, 58(4), 241-244 (2003-11-26)
A 36-year-old woman presented to the Emergency Room because of abdominal pain associated with hematuria and red blood blending to stool. On admission, the physical examination revealed abdominal tenderness and diffuse cutaneous hematoma. The laboratory findings showed abnormal clotting tests
S C Smolinske et al.
Pediatrics, 84(3), 490-494 (1989-09-01)
This prospective study was undertaken to determine the incidence, severity, time of onset, and duration of coagulopathy in children following accidental ingestion of long-acting anticoagulant rodenticides, often called "superwarfarins." Of 110 children, who ingested superwarfarins and in whom one or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service