Skip to Content
Merck
All Photos(2)

Documents

45871

Supelco

Disulfoton-sulfone

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19O4PS3
CAS Number:
Molecular Weight:
306.40
Beilstein:
1713538
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)SCCS(=O)(=O)CC

InChI

1S/C8H19O4PS3/c1-4-11-13(14,12-5-2)15-7-8-16(9,10)6-3/h4-8H2,1-3H3

InChI key

BKVJOVPVLOJPKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disulfoton-sulfone is a metabolite of the pesticide disulfoton, commonly used for different agricultural commodities.

Application

Disulfoton-sulfone may be used as an analytical reference standard for the quantification of the analyte in biological samples and wine samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agrochemicals Desk Reference, 1216(31), 5856-5867 (2009)
Agrochemicals Desk Reference, 1216(31), 5856-5867 (2009)
Rapid determination of disulfoton and its oxidative metabolites in human whole blood and urine using QuEChERS extraction and liquid chromatography-tandem mass spectrometry.
Usui K, et al.
Legal medicine, 14(6), 309-316 (2012)
Anastasios Economou et al.
Journal of chromatography. A, 1216(31), 5856-5867 (2009-06-30)
This work reports a new sensitive multi-residue liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for detection, confirmation and quantification of forty-six pesticides and transformation products belonging to different chemical classes in wines. The proposed method makes use of a solid-phase extraction
Jishi Wang et al.
Journal of chromatography. A, 1521, 10-18 (2017-09-25)
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service