Skip to Content
Merck
All Photos(1)

Documents

45655

Supelco

Resmethrin

PESTANAL®, analytical standard, mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26O3
CAS Number:
Molecular Weight:
338.44
Beilstein:
1626510
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

quality

mixture of isomers

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C(C)=C\C1C(C(=O)OCc2coc(Cc3ccccc3)c2)C1(C)C

InChI

1S/C22H26O3/c1-15(2)10-19-20(22(19,3)4)21(23)25-14-17-12-18(24-13-17)11-16-8-6-5-7-9-16/h5-10,12-13,19-20H,11,14H2,1-4H3

InChI key

VEMKTZHHVJILDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Resmethrin is a pyrethroid insecticide, used as a tool for the pest management.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Resmethrin may be used as a reference standard for the determination of resmethrin in insecticide formulations by gas chromatography.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pesticide exposures and respiratory health in general populations.
Ye M, et al.
Journal of environmental sciences (China), 51, 361-370 (2017)
Mark D Jankowski et al.
Environmental toxicology and chemistry, 36(12), 3376-3386 (2017-07-20)
West Nile virus (WNV) is maintained cryptically primarily in avian (passerine) populations, where it is transmitted by Culex spp. mosquitoes. Mosquito-control measures currently include physical activities to reduce mosquito-breeding sites and the application of mosquito larvicides or aerosolized insecticides to
Simultaneous determination of pynamin forte, resmethrin and piperonyl butoxide in insecticide formulations by gas chromatography.
Sura D J, et al.
Journal of Chromatography A, 314, 471-475 (1984)
Peter Key et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 40(4), 585-594 (2005-07-29)
This study investigated the toxicity of various concentrations of technical resmethrin and Scourge on adult and larval Palaemonetes pugio, a common grass shrimp species. Two types of tests were conducted for each of the resmethrin formulations using adult and larval
Derek W Gammon
Pest management science, 63(7), 625-627 (2007-06-05)
West Nile virus is becoming increasingly prevalent in the USA, causing fever, encephalitis, meningitis and many fatalities. Spread of the disease is reduced by controlling the mosquito vectors by a variety of means, including the use of pyrethroid insecticides, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service