Skip to Content
Merck
All Photos(1)

Key Documents

43791

Sigma-Aldrich

2,2′-Dithiodipyridine

≥99.0% (GC)

Synonym(s):

2,2′-Dipyridyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2S2
CAS Number:
Molecular Weight:
220.31
Beilstein:
154629
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99.0% (GC)

form

solid

mp

56-58 °C (lit.)
57-59 °C

storage temp.

2-8°C

SMILES string

S(Sc1ccccn1)c2ccccn2

InChI

1S/C10H8N2S2/c1-3-7-11-9(5-1)13-14-10-6-2-4-8-12-10/h1-8H

InChI key

HAXFWIACAGNFHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

For the determination of sulfhydryl groups in biological materials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of sulfhydryl groups with 2,2'- or 4,4'-dithiodipyridine.
D R Grassetti et al.
Archives of biochemistry and biophysics, 119(1), 41-49 (1967-03-01)
Rachel Lubong Sabado et al.
PloS one, 4(1), e4256-e4256 (2009-01-24)
The requirements for priming of HIV-specific T cell responses initially seen in infected individuals remain to be defined. Activation of T cell responses in lymph nodes requires cell-cell contact between T cells and DCs, which can give concurrent activation of
Sanae Haga et al.
Antioxidants & redox signaling, 11(10), 2563-2572 (2009-06-06)
Noninvasive evaluation of organ redox states provides invaluable information in many clinical settings. We evaluated a newly developed reduction/oxidation-sensitive green fluorescent protein (roGFP) probe that reports cellular redox potentials and their dynamic changes in live cells. On hypoxia/reoxygenation (H/R) of
T L Dutka et al.
The Journal of physiology, 589(Pt 9), 2181-2196 (2010-12-01)
S-Nitrosoglutathione (GSNO) is generated in muscle and may S-glutathionylate and/or S-nitrosylate various proteins involved in excitation–contraction (EC) coupling, such as Na+-K+-ATPases, voltage-sensors (VSs) and Ca2+ release channels (ryanodine receptors,RyRs), possibly changing their properties. Using mechanically skinned fibres from rat extensor
Rachel L Sabado et al.
European journal of immunology, 37(7), 1752-1763 (2007-05-31)
The outcome following HIV infection depends on the nature and durability of the HIV-specific T cell response induced initially. The activation of protective T cell responses depends upon dendritic cells (DC), antigen-presenting cells which have the capacity to process and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service