Skip to Content
Merck
All Photos(2)

Key Documents

42515

Supelco

Oleanolic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CC(C)(C)CC[C@@]1(CC[C@]3(C)C2=CC[C@]4([H])[C@@]5(C)CC[C@H](O)C(C)(C)[C@]5([H])CC[C@@]34C)C(O)=O

InChI

1S/C30H48O3/c1-25(2)14-16-30(24(32)33)17-15-28(6)19(20(30)18-25)8-9-22-27(5)12-11-23(31)26(3,4)21(27)10-13-29(22,28)7/h8,20-23,31H,9-18H2,1-7H3,(H,32,33)/t20-,21-,22+,23-,27-,28+,29+,30-/m0/s1

InChI key

MIJYXULNPSFWEK-GTOFXWBISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Triterpenoid isolated from medicinal plants. Antitumor and hepatoprotective agent. Oleanolic acid has therapeutic potential for neurodegenerative diseases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dick de Zeeuw et al.
The New England journal of medicine, 369(26), 2492-2503 (2013-11-12)
Although inhibitors of the renin-angiotensin-aldosterone system can slow the progression of diabetic kidney disease, the residual risk is high. Whether nuclear 1 factor (erythroid-derived 2)-related factor 2 activators further reduce this risk is unknown. We randomly assigned 2185 patients with
Liangfeng Fu et al.
Organic letters, 15(7), 1622-1625 (2013-03-28)
Bardoxolone methyl (2-cyano-3,12-dioxooleane-1,9(11)-dien-28-oic acid methyl ester; CDDO-Me) (1), a synthetic oleanane triterpenoid with highly potent anti-inflammatory activity (levels below 1 nM), has completed a successful phase I clinical trial for the treatment of cancer and a successful phase II trial
Min Song et al.
Journal of pharmaceutical and biomedical analysis, 40(1), 190-196 (2005-08-30)
A highly selective and sensitive HPLC-ESI-MS-MS method was developed for the determination of oleanolic acid in human plasma. The oleanolic acid and glycyrrhetinic acid (internal standard) were recovered from plasma with ethyl acetate liquid-liquid extraction. The organic extracts were dried
Fei Yu et al.
Journal of medicinal chemistry, 56(11), 4300-4319 (2013-05-15)
Development of hepatitis C virus (HCV) entry inhibitors represents an emerging approach that satisfies a tandem mechanism for use with other inhibitors in a multifaceted cocktail. By screening Chinese herbal extracts, oleanolic acid (OA) was found to display weak potency
Philippe Charlier et al.
Scientific reports, 3, 1296-1296 (2013-03-02)
During the Middle Ages, the partition of the cadaver of the elite members was a current practice, with highly technical treatment given to symbolic organs such as the heart. Considered mostly from a theoretical point of view, this notion of

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service