Skip to Content
Merck
All Photos(1)

Key Documents

38497

Millipore

Nitrate Reagent A

suitable for microbiology

Synonym(s):

1-Naphthylamine solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41171621
PubChem Substance ID:
NACRES:
NA.85

product line

BioChemika

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

acetic acid 5 N, 1000 mL
α-naphthylamine, 5 g

technique(s)

microbe id | metabolite detection: suitable

application(s)

agriculture
clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

Enterobacter spp.
Neisseria spp.
anaerobic bacteria
bacteria

SMILES string

Nc1c2c(ccc1)cccc2

InChI

1S/C10H9N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,11H2

InChI key

RUFPHBVGCFYCNW-UHFFFAOYSA-N

General description

α-Naphtylamine and sulfanilic acid are used for detection of nitrate reduction by bacteria. Organisms containing nitrate reductase reduce nitrate to nitrite, which forms a diazonium salt with sulfanilic acid and reacts with α-naphtylamine to form a red azo dye.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Time-dependent intensity changes of free fatty acids detected by matrix-assisted laser desorption and ionization time-of-flight mass spectrometry in the presence of 1,8-bis-(dimethylamino)naphthalene--a cautionary note.
Mandy Eibisch et al.
Rapid communications in mass spectrometry : RCM, 26(13), 1573-1576 (2012-05-29)
Xiaolin Zhang et al.
Inorganic chemistry, 51(4), 2325-2331 (2012-02-10)
A unique gadolinium complex, Nap-DO3A-Gd, comprising a naphthylamine luminescent moiety, a di-2-picolylamine (DPA) binding chelator, and a 1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid (DO3A) moiety has been designed and synthesized as a dual-functional probe for selective magnetic resonance imaging and fluorescent sensing of copper(II)
Arvydas Tamulis et al.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 43(1), 49-66 (2012-12-18)
The quantum mechanical self-assembly of two separate photoactive supramolecular systems with different photosynthetic centers was investigated by means of density functional theory methods. Quantum entangled energy transitions from one subsystem to the other and the assembly of logically controlled artificial
Zuofu Hu et al.
Optics letters, 37(15), 3072-3074 (2012-08-04)
An ultraviolet photodetector was fabricated based on Mg0.07Zn0.93O heterojunction. N, N'-bis (naphthalen-1-y1)-N, N'-bis(pheny) benzidine was selected as the hole transporting layer. I-V characteristic curves of the device were measured in the dark and under the illumination of 340 nm UV
Xiu-Li Liu et al.
Molecules (Basel, Switzerland), 18(2), 2183-2199 (2013-02-12)
Condensed tannins (CT) from purple prairie clover (PPC; Dalea purpurea Vent.) and sainfoin (SF; Onobrychis viciifolia) were assessed for anti-Escherichia coli activity by comparing their ability to react with proteins and liposome, cause cell aggregation, and alter outer membrane (OM)

Articles

Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service