Skip to Content
Merck
All Photos(1)

Key Documents

36137

Supelco

Isoproturon

PESTANAL®, analytical standard

Synonym(s):

3-(4-Isopropylphenyl)-1,1-dimethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O
CAS Number:
Molecular Weight:
206.28
Beilstein:
2214033
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)c1ccc(NC(=O)N(C)C)cc1

InChI

1S/C12H18N2O/c1-9(2)10-5-7-11(8-6-10)13-12(15)14(3)4/h5-9H,1-4H3,(H,13,15)

InChI key

PUIYMUZLKQOUOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoproturon is a phenylurea herbicide used to control ryegrass, silky bentgrass, and broadleaf species in spring and winter wheat.

Application

Isoproturon may be used as a reference standard for the determination of isoproturon in crops using high performance liquid chromatography (HPLC) with fluorescence detection combined with UV decomposition and post-column derivatization.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Degradation of the herbicide isoproturon by laccase-mediator systems.
Zeng S, et al.
Biochemical Engineering Journal, 119, 92-100 (2017)
Katja Knauer et al.
Ecotoxicology and environmental safety, 78, 178-183 (2011-12-14)
A mesocosm study with three photosystem-II inhibitors and an equipotent mixture was performed to address the value of functional and structural endpoints in evaluating the impact of herbicides on aquatic systems. The herbicides atrazine, diuron, and isoproturon were dosed in
Qing Ding et al.
Journal of hazardous materials, 190(1-3), 823-832 (2011-04-29)
Isoproturon is a selective herbicide belonging to the phenylurea family and widely used for pre- and post-emergence control of annual weeds. Soil amendments (e.g. organic compounds or dissolved organic matter) may affect environmental behavior and bioavailability of pesticides. However, whether
M Fernández-Pérez et al.
Journal of hazardous materials, 190(1-3), 794-801 (2011-04-26)
Important risk of groundwater pollution has been observed as a result of rapid leaching of highly soluble pesticides when used in agronomic practices as conventional formulations. This risk can be minimized through the application of the pesticide at a set
Julien Moeys et al.
Pest management science, 67(10), 1309-1319 (2011-05-14)
Sorption coefficients (the linear K(D) or the non-linear K(F) and N(F)) are critical parameters in models of pesticide transport to groundwater or surface water. In this work, a dataset of isoproturon sorption coefficients and corresponding soil properties (264 K(D) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service