Skip to Content
Merck
All Photos(1)

Key Documents

36510

Supelco

Buturon

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13ClN2O
CAS Number:
Molecular Weight:
236.70
Beilstein:
2941811
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C#C)N(C)C(=O)Nc1ccc(Cl)cc1

InChI

1S/C12H13ClN2O/c1-4-9(2)15(3)12(16)14-11-7-5-10(13)6-8-11/h1,5-9H,2-3H3,(H,14,16)

InChI key

BYYMILHAKOURNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Buturon belongs to the class of phenylurea herbicides, which finds applications in agriculture. Its mode of action involves the selective control of annual and perennial weeds via inhibition of their photosynthesis pathways.

Application

Buturon may be used as an analytical reference standard for the quantification of the analyte in aqueous samples using various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Studies on the administration of plant pesticides on the plant/soil system in lysimeters using 14C-marked substances with special reference to leaching behavior].
I Scheunert et al.
Schriftenreihe des Vereins fur Wasser-, Boden- und Lufthygiene, 68, 313-334 (1987-01-01)
Tso-Ying Chou et al.
Talanta, 80(2), 493-498 (2009-10-20)
Partitioned dispersive liquid-liquid microextraction (PDLLME), using THF as the dispersive solvent and dichloromethane as the extraction solvent, was utilized to isolate and concentrate phenylurea herbicides (PUHs) from aqueous samples. In PDLLME, a dispersive solvent should be able to partition in
Solid-phase microextraction coupled with high-performance liquid chromatography for the determination of phenylurea herbicides in aqueous samples.
Lin HH, et al.
Journal of Chromatography A, 1012(1), 57-66 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service