Skip to Content
Merck
All Photos(2)

Documents

D2425

Sigma-Aldrich

Diuron

≥98%

Synonym(s):

3-(3,4-Dichlorophenyl)-1,1-dimethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10Cl2N2O
CAS Number:
Molecular Weight:
233.09
Beilstein:
2215168
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

SMILES string

CN(C)C(=O)Nc1ccc(Cl)c(Cl)c1

InChI

1S/C9H10Cl2N2O/c1-13(2)9(14)12-6-3-4-7(10)8(11)5-6/h3-5H,1-2H3,(H,12,14)

InChI key

XMTQQYYKAHVGBJ-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Application

  • The Fabrication and Property Characterization of a Ho(2)YSbO(7)/Bi(2)MoO(6) Heterojunction Photocatalyst and the Application of the Photodegradation of Diuron under Visible Light Irradiation.: This research investigates a novel photocatalyst′s ability to degrade diuron under visible light, offering insights into sustainable environmental remediation technologies (Hao and Luan, 2024).

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Jahn et al.
Cell reports, 25(2), 478-486 (2018-10-12)
Cyanobacteria must balance separate demands for energy generation, carbon assimilation, and biomass synthesis. We used shotgun proteomics to investigate proteome allocation strategies in the model cyanobacterium Synechocystis sp. PCC 6803 as it adapted to light and inorganic carbon (Ci) limitation.
S Giacomazzi et al.
Chemosphere, 56(11), 1021-1032 (2004-07-28)
Diuron is a biologically active pollutant present in soil, water and sediments. A synthesis of literature data on its physicochemical properties, partitioning behaviour, abiotic and biotic transformations, toxicological and ecotoxicological impacts has been here performed. Data have shown that diuron
Lijin Tian et al.
Physical chemistry chemical physics : PCCP, 15(9), 3146-3154 (2013-01-24)
Photosystems I (PSI) and II (PSII) are two major pigment-protein complexes of photosynthetic organisms that function in series to convert sunlight energy into chemical energy. We have studied the picosecond fluorescence behaviour of the core of both photosystems in vivo
Tobias Wunder et al.
Planta, 237(2), 541-558 (2012-10-23)
Reversible phosphorylation of LHCII, the light-harvesting complex of photosystem II, controls its migration between the two photosystems (state transitions), and serves to adapt the photosynthetic machinery of plants and green algae to short-term changes in ambient light conditions. The thylakoid
Sheridan M Martin et al.
Journal of hazardous materials, 231-232, 70-78 (2012-07-17)
We studied the sorption-desorption behaviour of two herbicides (diuron and artrazine) in a soil rich in Fe and Al oxides (Ferrosol), either amended freshly with two different types of biochars or that contained biochars aged under field conditions. Standard batch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service