Skip to Content
Merck
All Photos(1)

Documents

36483

Supelco

Loganin

analytical standard

Synonym(s):

7-Hydroxy 6-desoxyverbenalin, Loganoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26O10
CAS Number:
Molecular Weight:
390.38
Beilstein:
55724
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]3[C@@H](C)[C@@H](O)C[C@H]13

InChI

1S/C17H26O10/c1-6-9(19)3-7-8(15(23)24-2)5-25-16(11(6)7)27-17-14(22)13(21)12(20)10(4-18)26-17/h5-7,9-14,16-22H,3-4H2,1-2H3/t6-,7+,9-,10+,11+,12+,13-,14+,16-,17-/m0/s1

InChI key

AMBQHHVBBHTQBF-UOUCRYGSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Iridoid glycoside isolated from various plants. Substrate for cytochrome P450

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo-Ram Jin et al.
Molecules (Basel, Switzerland), 24(9) (2019-05-06)
The aim of this study was to simplify and identify the contents of the herbal formula, HBX-5. This study was carried out to evaluate the therapeutic effects of HBX-6 in a mouse model of benign prostatic hyperplasia (BPH). Based on
Eun Ju Jeong et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(3-4), 317-321 (2011-10-11)
The methanolic extract of the fruits of Cornus officinalis S et Z. (Cornaceae) showed the significant neuroprotective activity against glutamate-induced toxicity in HT22 hippocampal cells. Chemical profile of n-BuOH fraction of the methanolic extract of C. officinalis fruits, which showed
Jatinder S Sangha et al.
PloS one, 7(8), e43990-e43990 (2012-09-07)
Liuwei Dihuang (LWDH), a classic Chinese medicinal formula, has been used to improve or restore declined functions related to aging and geriatric diseases, such as impaired mobility, vision, hearing, cognition and memory. Here, we report on the effect and possible
Chan Hum Park et al.
Toxicology, 290(1), 14-21 (2011-08-26)
Accumulating evidence indicates that uncontrolled diabetes leads to the progression of diabetic complications such as liver disorder. The present study was carried out to elucidate the protective role of loganin extracted from Corni Fructus against hepatic oxidative stress caused by
Wang-Lin Jiang et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(3-4), 217-222 (2011-10-08)
Connective tissue growth factor (CTGF) plays a pathogenic role in diabetic nephropathy (DN). Loganin, an iridoid glucoside compound was isolated from Cornus officinalis Sieb. et Zucc. This study was conducted to investigate the efficacy of loganin on DN and to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service