Skip to Content
Merck
All Photos(1)

Key Documents

33847

Supelco

Tylosin tartrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(C)=C\[C@@H]1CO[C@@H]4O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]4OC

InChI

1S/C46H77NO17.C4H6O6/c1-13-33-30(22-58-45-42(57-12)41(56-11)37(52)26(5)60-45)18-23(2)14-15-31(49)24(3)19-29(16-17-48)39(25(4)32(50)20-34(51)62-33)64-44-38(53)36(47(9)10)40(27(6)61-44)63-35-21-46(8,55)43(54)28(7)59-35;5-1(3(7)8)2(6)4(9)10/h14-15,17-18,24-30,32-33,35-45,50,52-55H,13,16,19-22H2,1-12H3;1-2,5-6H,(H,7,8)(H,9,10)/b15-14+,23-18+;/t24-,25+,26-,27-,28+,29+,30-,32-,33-,35+,36-,37-,38-,39-,40-,41-,42-,43+,44+,45-,46-;1-,2-/m11/s1

InChI key

ICVKYYINQHWDLM-KBEWXLTPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tylosin tartrate is a tylosin derivative and a medium spectrum antibiotic, which can find applications in veterinary medicine for the treatment of infections caused by most gram-positive bacteria, mycoplasmas, some gram-negative bacteria and chlamydia bacteria.[1] It can also be used as a feed additive in order to improve the weight gain and feed efficiency of cattle, poultry, turkey and swine.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tylosin is a mcrolide antibiotic commonly used in veterinary medicine. It has been used to study protein synthesis at the 50S ribosome, implant site abscess prevention in cattle, disposition kinetics and Mycoplasma gallisepticum and Mycoplasma meleagridis infections. It is recommended for use in cell culture applications at 8 mg/L.

Biochem/physiol Actions

Mode of Action: Tylosin Tartrate inhibits bacterial protein synthesis by binding to the 50S (L27 protein) ribosome.

Antimicrobial Spectrum: This product acts against gram-positive bacteria and mycoplasma.

Components

This product is a mixture of tylosin A, B, C, and D tartrates.

Caution

Stock solutions should be filter sterilized and stored at 2-8°C. These solutions are stable at 37°C for 3 days.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clara L Porfírio-Dias et al.
Journal of applied toxicology : JAT, 40(8), 1060-1066 (2020-03-10)
Andiroba (Carapa guianensis Aubl) is an Amazonian plant whose oil has been widely used in traditional medicine for various purposes, including anti-inflammation. Research reports indicate that the oil can confer antitumor activity due to the presence of fatty acids, which
Determination and correlation of solubility of tylosin tartrate in alcohol mixtures
Shen Y, et al.
The Journal of Chemical Thermodynamics, 80, 128-134 (2015)
N Gyémánt et al.
British journal of cancer, 103(2), 178-185 (2010-06-17)
The multidrug resistance (MDR) proteins are present in a majority of human tumours. Their activity is important to understand the chemotherapeutic failure. A search for MDR-reversing compounds was conducted among various Betti-base derivatives of tylosin. Here, we evaluate the in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service