Skip to Content
Merck
All Photos(1)

Key Documents

31727

Supelco

Lincomycin hydrochloride monohydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34N2O6S · HCl · H2O
CAS Number:
Molecular Weight:
461.01
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].[H]O[H].CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@H]2O

InChI

1S/C18H34N2O6S.ClH.H2O/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H;1H2/t9-,10-,11+,12-,13+,14-,15-,16-,18-;;/m1../s1

InChI key

LFZGYTBWUHCAKF-DCNJEFSFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside
Lincomycin hydrochloride monohydrate, an antibiotic used in treating infections caused by staphylococcal, streptococcal, and Bacteroides fragilis. It is produced by Streptomyces lincolnensis.

Application

Lincomycin HCl monohydrate may have been used as reference standard for determination of antibiotics in novel tablet formulation using HPLC.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Chih Tseng et al.
Plant molecular biology, 82(4-5), 375-392 (2013-05-07)
RNA editing is one of the post-transcriptional processes that commonly occur in plant plastids and mitochondria. In Arabidopsis, 34 C-to-U RNA editing events, affecting transcripts of 18 plastid genes, have been identified. Here, we examined the editing and expression of
Isaac M Hagenbuch et al.
Water research, 46(16), 5028-5036 (2012-07-24)
The role that antibiotics and other "emerging contaminants" play in shaping environmental microbial communities is of growing interest. The use of the prokaryotic metabolic inhibitors tylosin (T), lincomycin (L), and ciprofloxacin (C) in livestock and humans is both global and
Ulrich Schreiber et al.
Photosynthesis research, 114(3), 165-177 (2013-02-15)
A new type of multi-color PAM chlorophyll fluorometer (Schreiber et al. 2012) was applied for measurements of photodamage to photosystem II (PS II) in optically thin suspensions of Chlorella (200 μg Chl l(-1)) in the presence of 1 mM lincomycin.
Development and validation of a gradient HPLC method for the determination of clindamycin and related compounds in a novel tablet formulation.
Platzer DJ, White BA.
Journal of Pharmaceutical and Biomedical Analysis, 41(1), 84-88 (2006)
Phillip A Davis et al.
Photosynthesis research, 112(3), 153-161 (2012-06-15)
Plants use light to fix carbon through the process of photosynthesis but light also causes photoinhibition, by damaging photosystem II (PSII). Plants can usually adjust their rate of PSII repair to equal the rate of damage, but under stress conditions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service