Skip to Content
Merck
All Photos(3)

Key Documents

33615

Sigma-Aldrich

Starch

puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble

Synonym(s):

Starch, soluble

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

potato

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

grade

puriss. p.a.

form

powder

quality

soluble

impurities

≤0.7% subst. red. Fehling sol. (as maltose)

ign. residue

≤0.5% (as SO4)

loss

≤13% loss on drying, 105 °C

color

white to off-white

pH

6.0-7.5

suitability

passes test for test of sensitivity

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11+,12-/m1/s1

InChI key

GUBGYTABKSRVRQ-ASMJPISFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starch has been used as a test compound for preparing anhydroalditols like-1,5-anhydro-D-glucitol, 1,5-anhydro-D-mannitol, 2,5-anhydro-D-glucitol, 2,5-anhydro-D-mannitol and 1,4-anhydro-Dribitol.[1] It has also been used for preparing Ac medium for studying the fungal diversity in deep-sea hydrothermal ecosystems.[2]

Biochem/physiol Actions

Starch is a carbohydrate made up of glucose units joined by glycosidic bonds. Undigested part of potato starch is referred as Resistant starch (RS). Resistant starch, RS4 reduces the concentrations of total cholesterol and triglycerides in the serum as well as enhances the concentration of HDL-cholesterol in Wistar rats.[3]

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Anhydroalditols from Commodity Carbohydrates.
Letters in Organic Chemistry, 10 (2013)
Kacy Greenhalgh et al.
Cell reports, 27(5), 1621-1632 (2019-05-03)
By modulating the human gut microbiome, prebiotics and probiotics (combinations of which are called synbiotics) may be used to treat diseases such as colorectal cancer (CRC). Methodological limitations have prevented determining the potential combinatorial mechanisms of action of such regimens. We expanded
Reply to J.E. Galgani and G. Valentino.
Coby Eelderink et al.
The American journal of clinical nutrition, 97(4), 903-904 (2013-05-01)
Should insulin resistance degree be taken into account for assessment of glycemic index?
Jose E Galgani et al.
The American journal of clinical nutrition, 97(4), 902-903 (2013-03-22)
Qiang Wu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 928, 106-112 (2013-04-23)
The aqueous two-phase system (ATPS) with polyethylene glycol (PEG) and hydroxypropyl starch (HPS) was evaluated for the extraction of human immunoglobulin G (IgG) from human serum albumin (HSA). The partitions of IgG in PEG/sulphate, PEG/phosphate, PEG/Dextran and PEG/HPS ATPSs were

Questions

  1. Is this starch hydrolyzed?

    1 answer
    1. This starch is not hydrolyzed. Please see product S5651 for a hydrolyzed option:
      https://www.sigmaaldrich.com/product/sial/s5651.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service