Skip to Content
Merck
All Photos(1)

Documents

33578

Supelco

Microcystin RR-YR-LR solution

5 μg/mL in methanol (each), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

5 μg/mL in methanol (each)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

storage temp.

−20°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle D Onofrio et al.
Toxins, 12(5) (2020-05-18)
Many detection methods for phycotoxins, bioactive compounds produced by harmful algae, focus on one compound or a class of related compounds. Multiple harmful algal species often co-occur in the environment, however, emphasizing the need to analyze for the presence of
Tigist W Zewde et al.
Toxicon : official journal of the International Society on Toxinology, 153, 85-95 (2018-09-02)
Herein, we report the presence and concentrations of three most common variants of microcystin (MC-LR, -RR and -YR) in the liver and muscle tissues of wild Nile Tilapia (Oreochromis niloticus), Common Carp (Cyprinus carpio) and African Sharp Tooth Catfish (Clarias

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service